JKA7034
4arm-PEG20K
average Mn 20,000, Acrylate
Synonym(s):
4arm-PEG-Acrylate, 4arm-PEG-ACLT, Polyethylene glycol
About This Item
Recommended Products
Product Name
4arm-PEG20K-Acrylate, average Mn 20,000
form
solid
mol wt
average Mn 20,000
reaction suitability
reaction type: Polymerization Reactions
polymer architecture
shape: 4-arm
functionality: homofunctional
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service