Skip to Content
Merck
All Photos(1)

Key Documents

726621

Sigma-Aldrich

(R)-(+)-α-Methylbenzylamine

ChiPros®, produced by BASF, ≥99.0%

Synonym(s):

(R)-(+)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2410916
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.5 mmHg ( 20 °C)

Quality Level

Assay

≥99.0% (GC)
≥99.0%

form

liquid

optical purity

enantiomeric excess: ≥98.0%

refractive index

n20/D 1.526 (lit.)

bp

187-189 °C (lit.)

density

0.952 g/mL at 20 °C (lit.)

functional group

amine
phenyl

SMILES string

C[C@@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1

InChI key

RQEUFEKYXDPUSK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-α-Methylbenzylamine is a chiral amine.

Application

(R)-(+)-α-Methylbenzylamine may be used as a substrate to synthesize:
  • (S)-α-amino phosphonates
  • N-{(S)-[cyclohexan-(S)-2-ol]}-(R)-α-methylbenzyl amine and N-{(R)-[cyclohexan-(R)-2-ol]}-(R)-α-methylbenzyl amine
  • R-α-aminonitriles

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium perchlorate/diethylether catalyzed aminophosphonation of aldehydes.
Heydari A, et al.
Tetrahedron Letters, 39(37), 6729-6732 (1998)
New enantiomerically pure aminoalcohols from (R)-a-methylbenzylamine and cyclohexene oxide.
Barbaro P, et al.
Tetrahedron Asymmetry, 7(3), 843-850 (1996)
ChiPros Chiral Amines
Aldrich Chemfiles, 11(1) null
Karim Engelmark Cassimjee et al.
Organic & biomolecular chemistry, 10(28), 5466-5470 (2012-06-13)
For biocatalytic production of pharmaceutically important chiral amines the ω-transaminase enzymes have proven useful. Engineering of these enzymes has to some extent been accomplished by rational design, but mostly by directed evolution. By use of a homology model a key
Andrew M Kelly et al.
Nature protocols, 3(2), 215-219 (2008-02-16)
A three-component chiral derivatization protocol for determining the enantiopurity of chiral diols by (1)H NMR spectroscopic analysis is described here. The present approach involves the derivatization of 1,2- 1,3- and 1,4-diols with 2-formylphenylboronic acid and enantiopure alpha-methylbenzylamine. This method affords

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service