Skip to Content
Merck
All Photos(1)

Key Documents

77880

Sigma-Aldrich

(R)-(+)-α-Methylbenzylamine

purum, ≥98.0% (sum of enantiomers, GC)

Synonym(s):

(R)-(+)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2410916
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.5 mmHg ( 20 °C)

Quality Level

grade

purum

Assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D +30±2°, c = 10% in ethanol

optical purity

enantiomeric ratio: ≥97:3 (GC)

refractive index

n20/D 1.526 (lit.)
n20/D 1.528

bp

187-189 °C (lit.)

density

0.952 g/mL at 20 °C (lit.)

functional group

amine
phenyl

SMILES string

C[C@@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1

InChI key

RQEUFEKYXDPUSK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Standard chiral amine used for the resolution of acids. Use as chiral auxiliary

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

G. Knupp et al.
Chemische Berichte, 117, 2076-2076 (1984)
U. Schollkopf et al.
Angewandte Chemie (International Edition in English), 90, 136-136 (1978)
T. Volpe et al.
Tetrahedron Letters, 28, 2367-2367 (1987)
R.P. Polniaszek et al.
Tetrahedron Letters, 28, 4511-4511 (1987)
G. Revial et al.
Organic Syntheses, 70, 35-35 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service