Skip to Content
Merck
All Photos(3)

Key Documents

266981

Sigma-Aldrich

Nickel

powder, <50 μm, 99.7% trace metals basis

Synonym(s):

Nickel microparticles, Nickel micropowder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
EC Number:
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99.7% trace metals basis

form

powder

resistivity

6.97 μΩ-cm, 20°C

particle size

<50 μm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Testing Method

Average particle size determined by Fisher sub-sieve sizer

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ornella Valenti et al.
Cerebral cortex (New York, N.Y. : 1991), 19(3), 658-674 (2008-07-18)
The prefrontal cortex receives multiple inputs from the hippocampal complex, which are thought to drive memory-guided behavior. Moreover, dysfunctions of both regions have been repeatedly associated with several psychiatric disorders. Therefore, understanding the interconnections and modulatory interactions between these regions
A J Feilzer et al.
Nederlands tijdschrift voor tandheelkunde, 120(6), 335-341 (2013-07-19)
Orally applied metal alloys can cause undesirable physical effects. A distinction needs to be made in this respect between local and systemic reactions and toxic and immunological reactions. A case is presented which illustrates this problem. In this case, the
Marilia O S Seijo et al.
Journal of dental education, 77(5), 648-655 (2013-05-10)
Including students' perceptions in the educational process is considered a key component in monitoring the quality of academic programs. This study aimed to evaluate the concept of one's learning experience in endodontic teaching from the perspective of a group of
Michał Sarul et al.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 22(2), 253-260 (2013-05-28)
Cognizance of the mechanical properties of nickel-titanium archwires is necessary for the management of orthodontic therapy with fixed appliances. Acting on the periodontium with forces that are too heavy may lead to such complications as: pain, tooth root resorption and
Fariborz Amini et al.
International journal of orthodontics (Milwaukee, Wis.), 24(1), 43-44 (2013-06-05)
In this report, we present an uncomplicated approach to overcome lingual rolling of mandibular molars during orthodontic treatment, by using rectangular NiTi arch wire in twist fashion through the molar tube. The sequential steps of the twist manoeuvre on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service