Skip to Content
Merck
All Photos(1)

Key Documents

662844

Sigma-Aldrich

Allylcyclohexane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16
CAS Number:
Molecular Weight:
124.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.450

bp

148-149 °C/760 mmHg

density

0.803 g/mL at 25 °C

functional group

allyl

SMILES string

C=CCC1CCCCC1

InChI

1S/C9H16/c1-2-6-9-7-4-3-5-8-9/h2,9H,1,3-8H2

InChI key

KVOZXXSUSRZIKD-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service