M45008
Methylenecyclobutane
96%
Synonym(s):
exo-Methylenecyclobutane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C4H6(=CH2)
CAS Number:
Molecular Weight:
68.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
6.57 psi ( 20 °C)
Quality Level
Assay
96%
refractive index
n20/D 1.42 (lit.)
bp
42 °C/749 mmHg (lit.)
density
0.736 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C=C1CCC1
InChI
1S/C5H8/c1-5-3-2-4-5/h1-4H2
InChI key
QIRVGKYPAOQVNP-UHFFFAOYSA-N
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-29.2 °F - closed cup
Flash Point(C)
-34 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Guang Huan Shen et al.
Current medicinal chemistry, 27(35), 5918-5948 (2019-06-30)
The syntheses of acyclic nucleoside phosphonate (ANP) analogs linked with cyclic systems are described in the present review. The purpose of the review is to report the methodology of ANP analogs and to give an idea on the synthesis of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service