Skip to Content
Merck
All Photos(3)

Documents

529311

Sigma-Aldrich

Iron(III) oxide

≥99.995% trace metals basis

Synonym(s):

Ferric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe2O3
CAS Number:
Molecular Weight:
159.69
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.995% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

O=[Fe]O[Fe]=O

InChI

1S/2Fe.3O

InChI key

JEIPFZHSYJVQDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iron is present in the form of iron(III) oxide in almost all natural sources of water. It is the most common supplement of iron for algae. It is a highly stable n-type semiconductor, and possesses a band gap of 2.0 to 2.2eV. It is widely used in solar photoelectrolysis cells. Self-organized nanoporous iron(III) oxide (haematite) has been prepared. Orientation effects on nearly opaque single crystals of iron(III) oxide have been investigated by laser Raman spectroscopy.

Application

Iron(III) oxide may be used in the preparation of iron(III) oxide-hydrate gels.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The structure of iron (III) oxide-hydrate gels.
Van der Giessen AA.
J. Inorg. Nucl. Chem., 28(10), 2155-2159 (1966)
Light-induced reduction of natural iron (III) oxide and its relevance to phytoplankton.
Finden DAS, et al.
Nature, 309, 783-784 (1984)
Synthesis and photoelectrochemical properties of nanoporous iron (III) oxide by potentiostatic anodization.
Prakasam HE, et al.
Nanotechnology, 17(17), 4285-4285 (2006)
William B Ingler et al.
Journal of the American Chemical Society, 126(33), 10238-10239 (2004-08-19)
Stable zinc-doped iron(III) oxide thin films that exhibit p-type behavior were synthesized by spray pyrolytic deposition (SPD) on conducting indium-doped tin oxide-coated glass substrate. The highest photocurrent density of 1.1 mA/cm2 was observed at an illumination intensity of 40 mW/cm2
The single-crystal Raman spectra of nearly opaque materials. Iron (III) oxide and chromium (III) oxide.
Beattie IR and Gilson TR.
J. Chem. Soc. Sect. A, 980-986 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service