Skip to Content
Merck
All Photos(2)

Documents

636908

Sigma-Aldrich

Indium tin oxide coated glass slide, rectangular

surface resistivity 30-60 Ω/sq, slide

Synonym(s):

ITO, ITO coated slide, rectangular

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

surface resistivity

30-60 Ω/sq

L × W × thickness

75 mm × 25 mm × 1.1 mm

transmittance

>84%

refractive index

n20/D 1.517 (lit.)

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

Thickness of ITO coating is 300-600 Å.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leah V Sibener et al.
Cell, 174(3), 672-687 (2018-07-28)
TCR-signaling strength generally correlates with peptide-MHC binding affinity; however, exceptions exist. We find high-affinity, yet non-stimulatory, interactions occur with high frequency in the human T cell repertoire. Here, we studied human TCRs that are refractory to activation by pMHC ligands despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service