634409
Zinc titanate
nanopowder, <100 nm particle size (BET), 99% trace metals basis
Synonym(s):
Zinc titanium oxide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
Quality Level
Assay
99% trace metals basis
form
nanopowder
particle size
<100 nm (BET)
SMILES string
[Ti++].[O-]C([O-])=O
InChI
1S/3O.Ti.Zn/q;2*-1;;+2
InChI key
DJSKLKQRHSKRDZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Product of Engi-Mat Co.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Influence of nanostructured zinc titanate, zinc oxide or titanium dioxide thin film coated on fluorine doped tin oxide as working electrodes for dye-sensitized solar cell.
International Journal of Electrochemical Science, 7(8), 6787-6798 (2012)
Preparation of zinc titanate nanoparticles and their photocatalytic behaviors in the photodegradation of humic acid in water
Ceramics International, 38(5) (2012)
J. Alloy Compounds, 354, 303-303 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service