Skip to Content
Merck
All Photos(3)

Key Documents

338842

Sigma-Aldrich

Barium titanate(IV)

powder, <2 μm, 99.5% trace metals basis

Synonym(s):

Titanium barium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaTiO3
CAS Number:
Molecular Weight:
233.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.5% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: titanium

particle size

<2 μm

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Ti]([O-])=O

InChI

1S/Ba.3O.Ti/q+2;;2*-1;

InChI key

WNKMTAQXMLAYHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Barium titanate(IV) (BaTiO3) is a ceramic material that can be synthesized by reacting titanium oxide (TiO2) and barium salts in the presence of oleic acid as a stabilizing agent. It has a cubic crystal structure, which allows it to have ferroelectric, thermoelectric and piezoelectric properties. It forms a film with intense photoluminescence, which can be used for a wide range of electronic applications.

Application

BaTiO3 can be used to produce a nanocomposite for the fabrication of flexible nanogenerators. It can also be used as a filler material that can enhance the dielectric property of the composite.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of new polymer-BaTiO 3-composites with improved permittivity for embedded capacitors
Hanemann T, et al.
Microsystem technologies : sensors, actuators, systems integration, 17(2), 195-201 (2011)
Temperature treatment of nano-scaled barium titanate filler to improve the dielectric properties of high-k polymer based composites
Schumacher B, et al.
Microelectronic Engineering, 87(10), 1978-1983 (2010)
A general soft-chemistry route to perovskites and related materials: synthesis of BaTiO3, BaZrO3, and LiNbO3 nanoparticles
Niederberger M, et al.
Angewandte Chemie (International Edition in English), 43(17), 2270-2273 (2004)
Hydrothermal microwave: a new route to obtain photoluminescent crystalline BaTiO3 nanoparticles
Moreira ML, et al.
Chemistry of Materials, 20(16), 5381-5387 (2008)
Synthesis of barium titanate nanopowder using polymeric precursor method
Vinothini V, et al.
Ceramics International, 32(2), 99-103 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service