467634
Barium titanate(IV)
nanopowder (cubic crystalline phase), <100 nm particle size (BET), ≥99% trace metals basis
Synonym(s):
Titanium barium oxide
About This Item
Recommended Products
Quality Level
Assay
≥99% trace metals basis
form
nanopowder (cubic crystalline phase)
dielectric constant
150
reaction suitability
reagent type: catalyst
core: titanium
particle size
<100 nm (BET)
density
6.08 g/mL at 25 °C (lit.)
SMILES string
[Ba++].[O-][Ti]([O-])=O
InChI
1S/Ba.3O.Ti/q+2;;2*-1;
InChI key
WNKMTAQXMLAYHX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Physical form
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service