572357
Cerium(IV) oxide-gadolinium doped
nanopowder, contains 20 mol % gadolinium as dopant
Synonym(s):
Gadolinium doped ceria
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
nanopowder
Quality Level
contains
20 mol % gadolinium as dopant
composition
Ce0.8Gd0.2O2
reaction suitability
reagent type: catalyst
core: cerium
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
surface area
>100 m2/g
particle size
<100 nm
greener alternative category
, Enabling
SMILES string
[Gd].O=[Ce]=O
InChI
1S/Ce.Gd.2O
InChI key
ZBPTVGKVCOMWPA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Cerium(IV) oxide-gadolinium doped nanopowder (20 mol% gadolinium doped) is an electrolytic material with a high oxygen-ion conductivity, which operates at low temperatures. It can be prepared by a variety of methods such as sol-gel, wet chemical process, co-precipitation, hydrothermal and other combustion routes.
Gadolinium doped ceria may be prepared by cation complexation sol gel technique. XRD, specific surface area, morphology has been described in a study. Higher oxygen ion conductivity of GDC over zirconia based materials makes it a preferable choice for some components of SOFCs.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.
Application
Gadolinium doped ceria (GDC) may be used as an intermediate temperature electrolyte for solid oxide fuel cells (SOFCs) at elevated temperatures.
Packaging
Packaged in glass bottles
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis and properties of Gadolinium-doped ceria solid solutions for IT-SOFC electrolytes
International Journal of Hydrogen Energy, 33(13), 3480-3484 (2008)
Thermal expansion of Gd-doped ceria and reduced ceria.
Solid State Ionics, 132(3), 227-233 (2000)
Chemically-induced stresses in gadolinium-doped ceria solid oxide fuel cell electrolytes
Solid State Ionics, 95(3-4), 249-258 null
Microstructure and Grain-Boundary Effect on Electrical Properties of Gadolinium-Doped Ceria.
Journal of the American Ceramic Society. American Ceramic Society, 85(7), 1757-1762 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service