Skip to Content
Merck
All Photos(2)

Key Documents

734624

Sigma-Aldrich

Cerium(IV) oxide-samarium doped

greener alternative

<0.5 μm particle size, powder, contains 20 mol % samarium as dopant

Synonym(s):

SDC-20, SDC, Samarium-doped ceria

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

contains

20 mol % samarium as dopant

composition

Sm0.2Ce0.8O1.9

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

10-14 m2/g

particle size

0.1-0.5 μm (d50)
<0.5 μm

bulk density

1.787 g/mL

greener alternative category

SMILES string

[Sm].O=[Ce]=O

InChI

1S/Ce.2O.Sm

InChI key

AQGZISMLQKBRRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(IV) oxide-samarium doped powder (20% mol samarium doped) (SDC-20) is an electrolytic material with a high oxygen ion conductivity. Samarium ion acts as an aliovalent cation that can be incorporated into the host lattice to increase the oxygen vacancy concentration and improve the oxide ion conduction in cerium oxide. It can be synthesized by reverse strike co-precipitation technique.
Samarium doped ceria may be used as an electrolyte in solid oxide fuel cells (SOFCs). Electrical conductivity as a function of temperature of doped ceria was investigated in a study. Relation of dopant type, oxygen vacancy ordering and ionic conductivity was investigated by transmission electron microscopy and electron energy loss spectroscopy.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Cerium(IV) oxide-samarium doped can be used in the fabrication of solid oxide fuel cells (SOFCs), which can further be used as environmentally-friendly sources of alternative energy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugo J Avila-Paredes et al.
Physical chemistry chemical physics : PCCP, 11(38), 8580-8585 (2009-09-24)
We report a correlation between oxygen ionic conductivity and the enthalpy of formation of trivalent-doped ceria from the component binary oxides observed at relatively low temperatures (150-275 degrees C). The bulk conductivities of La-doped ceria samples identical to those previously
Stephan Immenschuh et al.
Transplantation, 99(1), 56-62 (2014-08-15)
Treatment of patients with antibody-mediated rejection (AMR) after kidney transplantation by rituximab and plasmapheresis is ambiguous. Because of its unknown efficiency and serious side effects, biomarkers, which are predictive for responsiveness to this treatment in AMR patients, are required. Twenty
Diego H Sanchez et al.
PLoS genetics, 10(11), e1004806-e1004806 (2014-11-21)
Epigenetic mechanisms suppress the transcription of transposons and DNA repeats; however, this suppression can be transiently released under prolonged heat stress. Here we show that the Arabidopsis thaliana imprinted gene SDC, which is silent during vegetative growth due to DNA
Jun Zhang et al.
Scientific reports, 4, 7082-7082 (2014-11-19)
In this work, redox-active Mn or Cr is introduced to the B site of redox stable perovskite Sr(0.95)Ti(0.9)Nb(0.1)O3.00 to create oxygen vacancies in situ after reduction for high-temperature CO2 electrolysis. Combined analysis using X-ray diffraction, X-ray photoelectron spectroscopy, transmission electron
Intermediate-temperature electrochemical performance of a polycrystalline PrBaCo2O5+ δ cathode on samarium-doped ceria electrolyte.
Chen D, et al.
Journal of Power Sources, 188(1), 96-105 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service