Skip to Content
Merck
All Photos(2)

Key Documents

572330

Sigma-Aldrich

Cerium(IV) oxide-gadolinium doped

greener alternative

nanopowder, contains 10 mol % gadolinium as dopant

Synonym(s):

Gadolinium doped ceria

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

Quality Level

contains

10 mol % gadolinium as dopant

composition

Ce0.9Gd0.1O2

reaction suitability

reagent type: catalyst
core: cerium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

>100 m2/g

particle size

5-10 nm (crystallite size)
<500 nm (secondary)

greener alternative category

SMILES string

[Gd].O=[Ce]=O

InChI

1S/Ce.Gd.2O

InChI key

ZBPTVGKVCOMWPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(IV) oxide-gadolinium doped nanopowder (10 mol% gadolinium doped) is an electrolytic material with high oxygen-ion conductivity, which operates at low temperatures. It can be prepared by a variety of methods such as sol-gel, wet chemical process, co-precipitation, hydrothermal and other combustion routes.
Gadolinium doped ceria may be prepared by cation complexation sol-gel technique. XRD, specific surface area, morphology has been described in a study. Higher oxygen ion conductivity of GDC over zirconia-based materials makes it a preferable choice for some components of SOFCs.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Gadolinium doped ceria (GDC) may be used as an intermediate temperature electrolyte for solid oxide fuel cells (SOFCs) at elevated temperatures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis and properties of Gadolinium-doped ceria solid solutions for IT-SOFC electrolytes
Fuentes RO and Baker RT
International Journal of Hydrogen Energy, 33(13), 3480-3484 (2008)
Thermal expansion of Gd-doped ceria and reduced ceria.
Hayashi H, et al.
Solid State Ionics, 132(3), 227-233 (2000)
Chemically-induced stresses in gadolinium-doped ceria solid oxide fuel cell electrolytes
Atkinson A, et al.
Solid State Ionics, 95(3-4), 249-258 null
Microstructure and Grain-Boundary Effect on Electrical Properties of Gadolinium-Doped Ceria.
Zhou XD, et al.
Journal of the American Ceramic Society. American Ceramic Society, 85(7), 1757-1762 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service