Skip to Content
Merck
All Photos(3)

Documents

563218

Sigma-Aldrich

Sodium nitrite

99.999% trace metals basis

Synonym(s):

Anti-Rust, Sodium nitrite (NaNO2 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

solid

autoignition temp.

914 °F

impurities

≤15.0 ppm Trace Metal Analysis

mp

271 °C (lit.)

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium nitrite (NaNO2) can be used for a variety of applications, such as:
  • corrosion inhibition
  • as an effective drug on cyanide poisoning
  • inhibiting the growth of micro-organisms

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Factors affecting the bacteriostatic action of sodium nitrite
Castellani AG and Niven Jr CF
Applied Microbiology, 3(3), 154-154 (1955)
Inhibition of copper corrosion in sodium nitrate solutions with nontoxic inhibitors
Scendo M
Corrosion Science, 50(6), 1584-1592 (2008)
Thiourea and substituted thioureas as corrosion inhibitors for aluminium in sodium nitrite solution
Subramanyam NC, et al.
Corrosion Science, 34(4), 563-571 (1993)
Jenna L Gordon et al.
Veterinary sciences, 7(2) (2020-04-29)
Neuroblastoma, the most common extracranial solid tumor in children, accounts for 15% of all pediatric cancer deaths. Pharmaceutical applications of S-Nitrosylation, which, under normal conditions is involved with a host of epigenetic and embryological development pathways, have exhibited great potential
Yen-Jung Chen et al.
Antioxidants & redox signaling, 31(2), 109-126 (2018-11-13)
Aims: The coordination of neurons to execute brain functions requires plenty of oxygen. Thus, it is not surprising that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service