Skip to Content
Merck
All Photos(5)

Key Documents

237213

Sigma-Aldrich

Sodium nitrite

ACS reagent, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Quality Level

Agency

suitable for EPA 300

Assay

≥97.0%

form

powder or crystals

autoignition temp.

914 °F

impurities

≤0.01% insolubles

mp

271 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.005%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium nitrite (NaNO2) is an inorganic compound that is commonly used as a reagent and catalyst in synthetic organic chemistry.

Application

Sodium nitrite can be used as a reagent in:      
  • The Sandmeyer reaction for converting amines into diazo derivatives, and nitration reaction.      
  • The oxidative C-C bond formation reaction in the presence of oxygen as the terminal oxidant.      
  • The oxidative carbonitration of alkenes in the presence of K2S2O8.

It can also be used as a nitrating agent in organic synthesis. For example, NaNO2 is used in direct C-H nitration, ipso-nitration, and nitration via transition metal-catalyzed cross-coupling reaction.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sania Bibi et al.
Mikrochimica acta, 186(9), 595-595 (2019-08-04)
A cyclic voltammetric method is described for the determination of nitrite by using a multiwalled carbon nanotube paste electrode (MWCNT) that was modified with chitosan-functionalized silver nanoparticles (Chit-AgNPs). The AgNPs were prepared by one step procedure using chitosan as stabilizing
Eagleson M.
Concise Encyclopedia Chemistry, 556-556 (1994)
P J McClure et al.
International journal of food microbiology, 14(1), 77-91 (1991-10-01)
An automated turbidimetric system using multiwelled plates was used to examine the effects of different combinations of NaCl (0.5-8.0% w/v), NaNO2 (0-400 micrograms/ml) pH (4.6-7.4) and temperature (5-30 degrees C) on the growth of Listeria monocytogenes in tryptone soya broth.
Saurabh Srivastava et al.
Scientific reports, 8(1), 11326-11326 (2018-07-29)
Neuroinflammation is recognized as one of the obligatory pathogenic features of neurodegenerative diseases including Alzheimer's, Parkinson's or prion diseases. In prion diseases, space and time correlations between deposition of disease-associated, pathogenic form of the prion protein or PrPSc and microglial-mediated
R Kokkola et al.
Scandinavian journal of immunology, 61(1), 1-9 (2005-01-13)
Abstract High-mobility group box chromosomal protein 1 (HMGB1) is a protein with both intranuclear functions and extracellular cytokine-like effects. In this report, we study possible candidate receptors for HMGB1 on macrophages (Mphi) and define pathways activated by HMGB1 binding. Bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service