Skip to Content
Merck
All Photos(2)

Key Documents

557684

Sigma-Aldrich

5-Bromo-2-thienylboronic acid

≥95%

Synonym(s):

5-Bromothiophene-2-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4BBrO2S
CAS Number:
Molecular Weight:
206.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

mp

95-100 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

OB(O)c1ccc(Br)s1

InChI

1S/C4H4BBrO2S/c6-4-2-1-3(9-4)5(7)8/h1-2,7-8H

InChI key

USJPOBDLWVCPGG-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki coupling reactions for the synthesis of benzotriazole-containing organic sensitizers and meso-Polyarylamide-BODIPY hybrids
  • Suzuki-Miyaura coupling for the synthesis of ratanhine
  • Microwave-assisted Sonogashira reactions for the synthesis of ethynylarylboronates

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Otsuka et al.
Materials (Basel, Switzerland), 13(20) (2020-10-15)
We have established a novel analytical method for solvent polarity on resin surface by combining the synthesis of fluorescent solvatochromic resin with optical waveguide spectrometry. The fluorescent solvatochromic resin was obtained via Suzuki-Miyaura cross-coupling between 4-iodobenzoic acid immobilized on Wang

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service