Skip to Content
Merck
All Photos(2)

Documents

436836

Sigma-Aldrich

2-Thienylboronic acid

≥95.0%

Synonym(s):

2-Thienylboric acid, 2-Thienylboronic acid, Thien-5-ylboronic acid, Thiophene-2-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5BO2S
CAS Number:
Molecular Weight:
127.96
Beilstein:
112375
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

solid

mp

138-140 °C (lit.)

storage temp.

2-8°C

SMILES string

OB(O)c1cccs1

InChI

1S/C4H5BO2S/c6-5(7)4-2-1-3-8-4/h1-3,6-7H

InChI key

ARYHTUPFQTUBBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used for
  • Palladium-catalyzed Suzuki-Miyaura cross-couplings
  • Alkylation, boration, coupling reaction, Suzuki coupling, and halogenation of fluorenyl bromide
  • Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer
  • Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters
  • Ligand-free Suzuki, Sonogashira, and Heck cross-coupling reactions
  • Copper-catalyzed nitration reactions
  • Geometry relaxation-induced Large Stokes shift in red-emitting borondipyrromethenes (BODIPY) and applications in fluorescent thiol probes

Reagent used in Preparation of
  • Photophysical properties of oxygen-containing polycyclic aromatic triptycenes
  • Donor unit for donor-acceptor-type polymers via N-alkylation, Suzuki coupling, and bromination
  • Aminopyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Ping Lu et al.
The Journal of organic chemistry, 77(8), 3700-3703 (2012-04-07)
The ligands associated with various Pd catalysts play a crucial role in determining the stereochemistry of cross-couplings between boronic acids and Z-alkenyl halides. A ligand on palladium has been found that leads to the desired products under mild conditions and
Synthesis of a novel donor unit for D-A-type polymers, 2,7-di(5-bromothiophen-2-yl)-10-hexadecyl-phenoxazine
Li, L.; et al.
Advanced Materials Research, 418-420 (2012)
Chain-growth catalyst transfer polycondensation of a conjugated alternating copolymer
Ono, R. J.; Bielawski, C. W.
Polymeric Materials: Science and Engineering Preprints (2012)
Yinghui Chen et al.
The Journal of organic chemistry, 77(5), 2192-2206 (2012-02-10)
2-Thienyl and 2,6-bisthienyl BODIPY derivatives (BS-SS and BS-DS) were prepared that show intense absorption (ε = 65000 M(-1) cm(-1) at 507 nm) and a large Stokes shift (96 nm) vs the small Stokes shift of typical BODIPY (<15 nm). Control
Synthesis of a novel fluorene derivative for polyfluorenes. 7,7'-Di(5-bromothiophen-2-yl)-9,9,9',9'-tetraoctyl-2,2'-bifluorene
Li, L.; et al.
Advanced Materials Research, 418-420 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service