Skip to Content
Merck
All Photos(1)

Key Documents

514055

Sigma-Aldrich

5-Formyl-2-thienylboronic acid

≥95.0%

Synonym(s):

5-Formyl-2-thiopheneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BO3S
CAS Number:
Molecular Weight:
155.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

132-135 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(s1)B(O)O

InChI

1S/C5H5BO3S/c7-3-4-1-2-5(10-4)6(8)9/h1-3,8-9H

InChI key

DEQOVKFWRPOPQP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond E Borg et al.
Photochemistry and photobiology, 95(1), 280-292 (2018-07-11)
The synthesis, photophysical and photoacoustic characterization for a series of nine π-extended quadrupolar curcumin dyes is presented. A systematic evaluation of the π-bridging unit including the p-phenyl, naphth-4-yl, thien-2-yl and hybrid 4-naphthathien-2-yl groups is presented. Furthermore, evaluation of the strongly
Haluk Dinçalp et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 202, 196-206 (2018-05-23)
Five isoindigo-based donor-acceptor-donor (D-A-D) type small molecules have been synthesized in order to investigate their intramolecular charge transfer characteristics. UV-vis absorption of these dyes exhibits a wide absorption band ranging from 300 to 650 nm with two distinct bands, giving the
Sara S M Fernandes et al.
ACS omega, 3(10), 12893-12904 (2018-11-10)
A series of push-pull heterocyclic N,N-diphenylhydrazones were prepared to study the effect of structural modifications (different π-spacers and electron-withdrawing groups) on the optical (linear and nonlinear) and electronic properties of the molecules. The photovoltaic response of dye-sensitized solar cells assembled
Gözde Murat Saltan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 372-381 (2017-08-02)
In an approach to develop efficient organic optoelectronic devices to be used in light-driven systems, a series of three thiophene linked benzimidazole conjugates were synthesized and characterized. The combination of two thiophene rings to a benzimidazole core decorated with different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service