Skip to Content
Merck
All Photos(2)

Documents

512117

Sigma-Aldrich

Benzo[b]thien-3-ylboronic acid

≥95.0%

Synonym(s):

Thianaphthene-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BO2S
CAS Number:
Molecular Weight:
178.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

225-230 °C (lit.)

SMILES string

OB(O)c1csc2ccccc12

InChI

1S/C8H7BO2S/c10-9(11)7-5-12-8-4-2-1-3-6(7)8/h1-5,10-11H

InChI key

QVANIYYVZZLQJP-UHFFFAOYSA-N

Application

Benzo[b]thien-3-ylboronic acid can be used:
  • To prepare thienyl substituted pyrimidine derivatives as potent antimycobacterial compounds.
  • To prepare 3-O-protected 17-heteroaryl-3-hydroxyestra-1,3,5,16-tetraene-16-carbaldehyde, which in turn is used for the synthesis of heteroarenes-annelated estranes.
  • As a substrate in the study of metal-free coupling reactions of allylic alcohols with heteroaryl boronic acids.
  • As a starting material for the preparation of thienyl based quinoline and pyridine ligands, which are further used to synthesize platinum complexes.

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, and structure-activity relationship for C (4) and/or C (5) thienyl substituted pyrimidines, as a new family of antimycobacterial compounds
Verbitskiy EV, et al.
European Journal of Medicinal Chemistry, 97, 225-234 (2015)
Heteroareno-annelated estranes by triene cyclization
Watanabe M, et al.
open chemistry, 4(3), 375-402 (2006)
Synthesis, characterization, and photophysical properties of bismetalated platinum complexes with benzothiophene ligands
Anderson CM, et al.
Journal of Organometallic Chemistry, 882, 10-17 (2019)
Metal-free allylation of electron-rich heteroaryl boronic acids with allylic alcohols
Li X, et al.
Tetrahedron, 72(15), 1873-1880 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service