Skip to Content
Merck
All Photos(2)

Key Documents

518158

Sigma-Aldrich

Iron(II,III) oxide

99.99% trace metals basis

Synonym(s):

Ferrosoferric oxide, Iron oxide black, Magnetite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe3O4
CAS Number:
Molecular Weight:
231.53
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

mp

1538 °C (lit.)

density

4.8-5.1 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Fe].O=[Fe]O[Fe]=O

InChI

1S/3Fe.4O

InChI key

SZVJSHCCFOBDDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iron(II,III) oxide, also known as ferric ferrous oxide, with a purity of 99.99% trace metals basis, exhibits unique magnetic, electrical, and catalytic properties. It has density of 4.8-5.1 g/mL at 25 °C and melting point of 1538 °C , which indicates thermal stability and mechanical strength. In the field of biomedical applications, it has shown promise in magnetic resonance imaging (MRI) contrast agents and drug delivery systems due to its magnetic properties and biocompatibility. In the semiconductor industry, iron(II,III) oxide has been investigated for its potential use in spintronics and magnetic tunnel junctions, where its magnetic properties can be utilized for information storage and processing. Additionally, it is being explored for use as an anode material in lithium-ion batteries, offering high theoretical capacity and low cost due to its abundance.

Application


  • Achieving Excellent Dielectric and Energy Storage Performance in Core-Double-Shell-Structured Polyetherimide Nanocomposites.: This study explores the development of polyetherimide nanocomposites incorporating Iron(II,III) oxide for enhanced dielectric properties and energy storage capabilities, showing potential for advanced electrical applications (Yuan et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongxing Hu et al.
Journal of the American Chemical Society, 135(6), 2213-2221 (2013-01-26)
Controlled assembly of nanoparticles into asymmetric configurations is of great interest due to their novel properties and promising applications. In this Article, we report a generic strategy for the synthesis of dimer nanoclusters and asymmetric nanoassemblies by using magnetic colloidal
Marina I Siponen et al.
Nature, 502(7473), 681-684 (2013-10-08)
Magnetotactic bacteria align along the Earth's magnetic field using an organelle called the magnetosome, a biomineralized magnetite (Fe(II)Fe(III)2O4) or greigite (Fe(II)Fe(III)2S4) crystal embedded in a lipid vesicle. Although the need for both iron(II) and iron(III) is clear, little is known
Jens Baumgartner et al.
Nature materials, 12(4), 310-314 (2013-02-05)
The formation of crystalline materials from solution is usually described by the nucleation and growth theory, where atoms or molecules are assumed to assemble directly from solution. For numerous systems, the formation of the thermodynamically stable crystalline phase is additionally
A Aranda et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 954-963 (2013-01-30)
No consensus exists on how to address possible toxicity of nanomaterials as they interfere with most in vitro screening tests based on colorimetric and fluorimetric probes such as the dichloro-dihydro-fluorescein diacetate (DCFH-DA) assay for detection of oxidative species. In the
Leyong Zeng et al.
Nanoscale, 5(5), 2107-2113 (2013-02-06)
Multifunctional Fe(3)O(4)-TiO(2) nanocomposites with Janus structure for magnetic resonance imaging (MRI) and potential photodynamic therapy (PDT) were synthesized, in which Fe(3)O(4) was used as a MRI contrast agent and TiO(2) as an inorganic photosensitizer for PDT. Their morphology, structure, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service