Skip to Content
Merck
All Photos(3)

Documents

412287

Sigma-Aldrich

Pyromellitic dianhydride

97%

Synonym(s):

Benzene-1,2,4,5-tetracarboxylic dianhydride, PMDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H2O6
CAS Number:
Molecular Weight:
218.12
Beilstein:
213583
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

bp

397-400 °C (lit.)

mp

283-286 °C (lit.)

SMILES string

O=C1OC(=O)c2cc3C(=O)OC(=O)c3cc12

InChI

1S/C10H2O6/c11-7-3-1-4-6(10(14)16-8(4)12)2-5(3)9(13)15-7/h1-2H

InChI key

ANSXAPJVJOKRDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyromellitic dianhydride is a dianhydride class of monomers commonly used in the preparation of polyimides polymers which are used in aerospace, electronics, and automotive industries because of their excellent thermal stability, high mechanical strength, and inherent flame resistance. It is also used in the synthesis of thermoplastic polymers such as polyesters and polyethers, as well as plasticizers and epoxy resins. Additionally, PMDA serves as a curing agent for epoxy resins. Epoxy resins are widely used in adhesives, coatings, composites, and electrical insulation materials.

Pyromellitic dianhydride (PMDA) is an acidic anhydride that can be used as a repair agent and as a chain extender in the formation of polyethylene terephthalate (PET) based chain extensions. It is mainly used in the production of thermoplastics and other coating applications.

Application

PMDA can be used as:
  • A monomer to synthesize aromatic polyimides with excellent thermo-mechanical and chemical properties. These polymers find the applications in automotive and electronic industries.
  • A capping agent in the development of siloxane-based hybrid materials for potential usage in organic electronics.
  • A monomer in the preparation and modification of thin film composite membranes, which are used in water purification, gas separation, and biomedical devices.
  • A monomer in the synthesis of pyromellitic diimide-based copolymers as stable cathode active materials for lithium and sodium-ion batteries.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

716.0 °F - closed cup

Flash Point(C)

380 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia Anceschi et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-12)
Cyclodextrin (CD)-based polymers are known to efficiently form molecular inclusion complexes with various organic and inorganic guest compounds. In addition, they also have a great potential as metal complexes because deprotonated hydroxyls can strongly bind metal ions under alkaline conditions.
Shadpour Mallakpour et al.
Amino acids, 43(4), 1605-1613 (2012-02-14)
In the present work, several novel optically active nanostructure poly(amide-imide)s (PAI)s were synthesized via step-growth polymerization reaction of chiral diacids based on pyromellitic dianhydride-derived dicarboxylic acids containing different natural amino acids such as L-alanine, S-valine, L-leucine, L-isoleucine, L-methionine, and L-phenylalanine
High-refractive-index thin films prepared from aminoalkoxysilane-capped pyromellitic dianhydride-titania hybrid materials
Chang C and Chen W
Journal of Polymer Science Part A: Polymer Chemistry, 39(19), 3419-3427 (2001)
Jun-Xia Yu et al.
Bioresource technology, 110, 160-166 (2012-02-22)
Magnetic pyromellitic dianhydride (PMDA) modified sugarcane bagasse (SCB) was prepared by a situ co-precipitation method. Results showed that the magnetic modified SCB could be recycled easily by an applied magnetic field. Adsorption capacities of the magnetic sorbent for cationic dyes:
Yiwu Liu et al.
Polymers, 12(9) (2020-09-12)
A high-barrier polyimide (2,7-CPI) was synthesized through the polymerization of pyromellitic dianhydride (PMDA) and a novel diamine (2,7-CDA) containing carbazole moiety. The synthesized diamine and polyimide were fully characterized by elemental analyses, FTIR and NMR. The 2,7-CPI displays very attractive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service