Skip to Content
Merck
All Photos(1)

Key Documents

J2500

Sigma-Aldrich

(±)-Jasmonic acid

BioReagent, suitable for plant cell culture, liquid

Synonym(s):

(±)-1α,2β-3-Oxo-2-(cis-2-pentenyl)cyclopentaneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
form:
liquid

product line

BioReagent

Quality Level

Assay

≥97% (TLC)

form

liquid

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

SMILES string

CC\C=C/C[C@H]1[C@@H](CCC1=O)CC(O)=O

InChI

1S/C12H18O3/c1-2-3-4-5-10-9(8-12(14)15)6-7-11(10)13/h3-4,9-10H,2,5-8H2,1H3,(H,14,15)/b4-3-/t9-,10-/m0/s1

InChI key

ZNJFBWYDHIGLCU-CMIOBCHKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Jasmonic acid (JA) is a plant hormone derived from linolenic acid. Jasmonic acid is involved in plant cell signaling responses to abiotic and biotic stress. JA is involved in the regulation of processes that include growth inhibition, senescence, flower development, leaf abscission and tendril coiling. JA is also involved in wound healing of plants and systemic acquired resistance to attack by insects.

Preparation Note

Other Notes

Note: This product is a clear, colorless to very light yellow oil that adheres to the surface of the vial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélie Gfeller et al.
Science signaling, 3(109), cm3-cm3 (2010-02-18)
Plants possess a family of potent fatty acid-derived wound-response and developmental regulators: the jasmonates. These compounds are derived from the tri-unsaturated fatty acids alpha-linolenic acid (18:3) and, in plants such as Arabidopsis thaliana and tomato, 7(Z)-, 10(Z)-, and 13(Z)-hexadecatrienoic acid
Alejandra Chacón-López et al.
Plant signaling & behavior, 6(3), 382-392 (2011-03-04)
Plants are exposed to several biotic and abiotic stresses. A common environmental stress that plants have to face both in natural and agricultural ecosystems that impacts both its growth and development is low phosphate (Pi) availability. There has been an
Kyoungwon Cho et al.
Journal of proteome research, 6(9), 3581-3603 (2007-08-23)
Two global approaches were applied to develop an inventory of differentially expressed proteins and genes in rice (cv. Nipponbare) seedling grown on Murashige and Skoog medium with and without jasmonic acid (JA). JA significantly reduced the growth of shoot, root
Yu Hua Wang et al.
Plant signaling & behavior, 6(4), 494-500 (2011-03-17)
Plant growth and development is influenced by mutual interactions among plant hormones. The five classical plant hormones are auxins, cytokinins, gibberellins, abscisic acid and ethylene. They are small diffusible molecules that easily penetrate between cells. In addition, newer classes of
Longfei Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(13) (2021-03-28)
Polyploidy is a prominent feature for genome evolution in many animals and all flowering plants. Plant polyploids often show enhanced fitness in diverse and extreme environments, but the molecular basis for this remains elusive. Soil salinity presents challenges for many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service