Skip to Content
Merck
All Photos(3)

Key Documents

476706

Sigma-Aldrich

Polyaniline (emeraldine base)

average Mw ~10,000

Synonym(s):

Emeraldine base polyaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw ~10,000

Quality Level

solubility

m-cresol: soluble
DMAC: soluble
DMF: soluble
DMSO: soluble
NMP: soluble

InChI

1S/C3H4BrN3.ClH/c1-2-3(4)6-7-5-2;/h1H3,(H,5,6,7);1H

InChI key

LYCWKZFKTMHXQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyaniline (PAni) emeraldine salt can be prepared by protonation of PAni emeraldine base (PAni-EB). PAni-EB forms at pH>7. It′s mechanism against corrosive agents has been explained in a study.
Polyaniline (emeraldine base) (PANI) is an intrinsic conductive polymer which consists of a ring structure attached with nitrogen substituted derivatives. It has an emeraldine type of oxidation state that exists in the form of its base. It exhibits high corrosion resistance and can be used in the development of electrochemical devices.

Application

Polyaniline (PANI) forms a conjugating system that can be used for a variety of semiconducting applications. It is also widely used as protective coatings and hole transporting layers (HTLs) for organic electronics and chemical sensing.
The polymer acts as an anticorrosive additive. Epoxy paints may be modified using PAni-EB.

Features and Benefits

Undoped form of conducting polymer (non-conductive).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticorrosion performances of epoxy coatings modified with polyaniline: A comparison between the emeraldine base and salt forms
Armelin E, et al.
Progress in Organic Coatings, 65(1), 88-93 (2009)
Corrosion protection of iron/steel by emeraldine base polyaniline: an X-ray photoelectron spectroscopy study.
Fahlman M, et al.
Synthetic Metals, 85(1-3), 1323-1326 (1997)
Study of epoxy and alkyd coatings modified with emeraldine base form of polyaniline.
Armelin E, et al.
Progress in Organic Coatings, 58(4), 316-322 (2007)
Suitability of emeraldine base polyaniline-PVA composite film for carbon dioxide sensing.
Irimia-Vladu M and Fergus JW
Synthetic Metals, 156(21-24), 1401-1407 (2006)
Application of polyaniline (emeraldine base, EB) in polymer light-emitting devices.
Wang HL, et al.
Synthetic Metals, 78(1), 33-37 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service