Skip to Content
Merck
All Photos(1)

Key Documents

458775

Sigma-Aldrich

(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO3
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D +97°, c = 1 in ethanol

optical purity

ee: 99% (HPLC)

mp

44-46 °C (lit.)

functional group

phenyl

SMILES string

CCC(=O)N1[C@H](COC1=O)Cc2ccccc2

InChI

1S/C13H15NO3/c1-2-12(15)14-11(9-17-13(14)16)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3/t11-/m0/s1

InChI key

WHOBYFHKONUTMW-NSHDSACASA-N

Application

Versatile chiral auxiliary for asymmetric synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ager, D.J., et al.
Aldrichimica Acta, 30, 3-3 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service