530689
Polyaniline (emeraldine base)
average Mw ~65,000
Synonym(s):
Emeraldine base polyaniline
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
mol wt
average Mw ~65,000
Quality Level
mp
>350 °C
solubility
m-cresol: soluble
DMAC: soluble
DMF: soluble
DMSO: soluble
NMP: soluble
λmax
331 nm
InChI
1S/C3H4BrN3.ClH/c1-2-3(4)6-7-5-2;/h1H3,(H,5,6,7);1H
InChI key
LYCWKZFKTMHXQA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Polyaniline (PAni) emeraldine salt can be prepared by protonation of PAni emeraldine base (PAni-EB). PAni-EB forms at pH>7. Its mechanism against corrosive agents has been explained in a study.
Polyaniline (emeraldine base) (PANI) is a conductive polymer which consists of a ring structure attached with nitrogen substituted derivatives. It has an emeraldine type oxidation state that exists in the form of its base. It can be prepared by deprotonation of emeraldine hydrochloric acid and ammonium hydroxide. It is mainly used in the development of electrochemical devices.
Application
PANI forms a conjugating polymeric system that can be used for a variety of semiconducting applications. It can be used in the fabrication of a variety of devices which include fuel cells, light emitting diodes supercapacitors, chemical sensors and rechargeable batteries.
PAni-EB/Polyacrylonitrile (PAN) composite nanofibers were fabricated. The effect of the concentration of dopant and solvent on the morphology, conductivity and mechanical properties of the composites were studied. Preparation of PAni-EB/PAN nanoweb has also been reported.
Features and Benefits
Undoped form of conducting polymer (non-conductive).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Carbon nanotube/polyaniline composite as anode material for microbial fuel cells
Journal of Power Sources, 170(1), 79-84 (2007)
Application of polyaniline (emeraldine base, EB) in polymer light-emitting devices.
Synthetic Metals, 78(1), 33-37 (1996)
Supercapacitors based on flexible graphene/polyaniline nanofiber composite films
ACS Nano, 4(4), 1963-1970 (2010)
A Polyaniline-Coated Sulfur/Carbon Composite with an Enhanced High-Rate Capability as a Cathode Material for Lithium/Sulfur Batteries
Advanced Energy Materials, 2(10), 1238-1245 (2012)
Polyanilines: a novel class of conducting polymers.
Faraday Discussions of the Chemical Society, 88(1-3), 317-332 (1989)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service