Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyaniline (emeraldine base)
Synonym(s):
Emeraldine base polyaniline
CAS Number:
25233-30-1
Products
Brand
Sigma-Aldrich (6)
Melting Point (°C)
Quality Segment
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
576379
Polyaniline (emeraldine base),
average M
w
~100,000
Expand
View Pricing
556459
Polyaniline (emeraldine base),
average M
w
~5,000
Expand
View Pricing
556386
Polyaniline (emeraldine base),
average M
w
~50,000
Expand
View Pricing
556378
Polyaniline (emeraldine base),
average M
w
~20,000
Expand
View Pricing
476706
Polyaniline (emeraldine base),
average M
w
~10,000
Expand
View Pricing
530689
Polyaniline (emeraldine base),
average M
w
~65,000
Expand
View Pricing