Skip to Content
Merck
All Photos(2)

Documents

474037

Sigma-Aldrich

Poly(butyl methacrylate-co-methyl methacrylate)

average Mw ~150,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)[CO2(CH2)3CH3]]X[CH2C(CH3)(CO2CH3)]Y
CAS Number:
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

mol wt

average Mw ~150,000

reduced viscosity

50-60 cm3/g(20 °C)(lit.)

acid number

1‑2 mg KOH/g

transition temp

Tg (DSC) 52 °C (onset)

solubility

esters, ketones, glycol ethers and aromatics: soluble
ethanol and water: insoluble

density

1.08 g/mL at 25 °C

SMILES string

COC(=O)C(C)=C.CCCCOC(=O)C(C)=C

InChI

1S/C8H14O2.C5H8O2/c1-4-5-6-10-8(9)7(2)3;1-4(2)5(6)7-3/h2,4-6H2,1,3H3;1H2,2-3H3

InChI key

WHLPIOPUASGRQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Binding agent for printing inks and foil, hardboard and ceramic lacquers.
Poly(butyl methacrylate-co-methyl methacrylate) (P(BMA-co-MMA) ) may be used to synthesize porous interpenetrating polymer network (IPN) with low-density polymers. These IPN structures are applicable as membranes for separation of liquid mixtures due to their high mechanical strength and selectivity. P(BMA-co-MMA) particles may be used to encapsulate single non-aggregated semiconducting quantum dots (QDs) for size enlargement.These hybrid organic/inorganic nanostructures may be used as single photon sources.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphology of crosslinked poly (butyl methacrylate-co-methyl methacrylate) porous membranes.
Lim B Y and Kim S C
Journal of Membrane Science , 209(1), 293-307 (2002)
Encapsulating of single quantum dots into polymer particles.
Gao Y, et al.
Colloid and Polymer Science, 286(11), 1329-1334 (2008)
Synthesis, electron irradiation modification and characterization of polyethylene/poly (butyl methacrylate?co?methyl methacrylate) interpenetrating polymer network.
Hu J, et al.
Polymers For Advanced Technologies, 9(10?11), 746-751 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service