Skip to Content
Merck
All Photos(1)

Key Documents

252034

Sigma-Aldrich

1,1′,3,3,3′,3′-Hexamethylindotricarbocyanine iodide

97%

Synonym(s):

2-[7-(1,3-Dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)-1,3,5-heptatrienyl]-1,3,3-trimethyl-3H-indolium iodide, HITC iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H33IN2
CAS Number:
Molecular Weight:
536.49
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

impurities

2% ethyl alcohol

mp

198 °C (dec.) (lit.)

λmax

740 nm

SMILES string

[I-].CN1c2ccccc2C(C)(C)\C1=C\C=C\C=C\C=C\C3=[N+](C)c4ccccc4C3(C)C

InChI

1S/C29H33N2.HI/c1-28(2)22-16-12-14-18-24(22)30(5)26(28)20-10-8-7-9-11-21-27-29(3,4)23-17-13-15-19-25(23)31(27)6;/h7-21H,1-6H3;1H/q+1;/p-1

InChI key

JKXWXYURKUEZHV-UHFFFAOYSA-M

Application

Suitable as laser dye

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruoyang Yao et al.
Light, science & applications, 8, 26-26 (2019-03-12)
Macroscopic fluorescence lifetime imaging (MFLI) via compressed sensed (CS) measurements enables efficient and accurate quantification of molecular interactions in vivo over a large field of view (FOV). However, the current data-processing workflow is slow, complex and performs poorly under photon-starved
Jason T Smith et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(48), 24019-24030 (2019-11-14)
Fluorescence lifetime imaging (FLI) provides unique quantitative information in biomedical and molecular biology studies but relies on complex data-fitting techniques to derive the quantities of interest. Herein, we propose a fit-free approach in FLI image formation that is based on
Judith Weber et al.
Cancer research, 79(20), 5407-5417 (2019-08-29)
Reactive oxygen species play an important role in cancer, however, their promiscuous reactivity, low abundance, and short-lived nature limit our ability to study them in real time in living subjects with conventional noninvasive imaging methods. Photoacoustic imaging is an emerging
M Ochoa et al.
Biomedical optics express, 11(10), 5401-5424 (2020-11-06)
Single pixel imaging frameworks facilitate the acquisition of high-dimensional optical data in biological applications with photon starved conditions. However, they are still limited to slow acquisition times and low pixel resolution. Herein, we propose a convolutional neural network for fluorescence
Hong Vu et al.
Bioorganic & medicinal chemistry, 27(9), 1855-1862 (2019-03-27)
Many imaging probes have been developed for a wide variety of imaging modalities. However, no optical imaging probe could be utilized for both microscopic and whole animal imaging. To fill the gap, the dual-wavelength fluorescent imaging nanoprobe was developed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service