Skip to Content
Merck
All Photos(2)

Key Documents

364681

Sigma-Aldrich

2-(Trimethylsilyl)ethanethiol

95%

Synonym(s):

(2-Mercaptoethyl)trimethylsilane, BEST

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CH2SH
CAS Number:
Molecular Weight:
134.32
Beilstein:
1732126
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

144-146 °C (lit.)
62-63 °C/43 mmHg (lit.)

density

0.839 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)CCS

InChI

1S/C5H14SSi/c1-7(2,3)5-4-6/h6H,4-5H2,1-3H3

InChI key

BCOLNMGFOWHFNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The 2-(trimethylsilyl) ethyl sulfur group in synthesis.
Chambert S, et al.
Synthesis, 16, 2319-2334 (2002)
Annika Karch et al.
Journal of clinical epidemiology, 78, 73-82 (2016-04-25)
To investigate how choice of gold standard biases estimates of sensitivity and specificity in studies reassessing the diagnostic accuracy of biomarkers that are already part of a lifetime composite gold standard (CGS). We performed a simulation study based on the
Ana Karen López-Contreras et al.
Antioxidants (Basel, Switzerland), 9(9) (2020-09-24)
Diabetic retinopathy is one of the leading causes of visual impairment and morbidity worldwide, being the number one cause of blindness in people between 27 and 75 years old. It is estimated that ~191 million people will be diagnosed with
Chi Zhang et al.
Molecular biology and evolution, 35(2), 504-517 (2017-12-09)
Reticulate species evolution, such as hybridization or introgression, is relatively common in nature. In the presence of reticulation, species relationships can be captured by a rooted phylogenetic network, and orthologous gene evolution can be modeled as bifurcating gene trees embedded
Michelle A Gordy et al.
Environmental health : a global access science source, 17(1), 73-73 (2018-10-26)
Cercarial dermatitis, colloquially "swimmer's itch", is a rash contracted in natural bodies of water, when people are exposed to skin-penetrating, larval flatworm parasites of the family Schistosomatidae, that emerge from aquatic snails. Swimmer's itch is a globally-distributed, allergic condition, of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service