Skip to Content
Merck
All Photos(3)

Key Documents

417556

Sigma-Aldrich

4-Fluorophenylboronic acid

≥95%

Synonym(s):

(4-Fluorophenyl)boric acid, (4-Fluorophenyl)dihydroxyborane, (4-Fluorophenyl)dihydroxyboron, (p-Fluorophenyl)boric acid, 4-Fluorobenzeneboronic acid, p-Fluorobenzylboronic acid, p-Fluorophenylboronic acid, NSC 142683

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4B(OH)2
CAS Number:
Molecular Weight:
139.92
Beilstein:
2829653
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

262-265 °C (lit.)

functional group

fluoro

SMILES string

OB(O)c1ccc(F)cc1

InChI

1S/C6H6BFO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4,9-10H

InChI key

LBUNNMJLXWQQBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluorophenylboronic acid can be used as a reactant in coupling reactions with arenediazonium tetrafluoroborates, iodonium salts, and iodanes. It is also used to make novel biologically active terphenyls.

It can also be used as a reactant in:
  • Suzuki coupling using microwave and triton B catalyst.
  • Pd-catalyzed direct arylation of pyrazoles with phenylboronic acids.
  • Mizoroki-Heck and Suzuki-Miyaura coupling reactions catalyzed by palladium nanoparticles.
  • Cu-catalyzed Petasis reactions.
  • Tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence.
  • Ruthenium catalyzed direct arylation.
  • Rh-catalyzed asymmetric conjugate additions.
  • Ligand-free copper-catalyzed coupling of nitro arenes with arylboronic acids.
  • Regioselective arylation and alkynylation by Suzuki-Miyaura and Sonogashira cross-coupling reactions.
  • Suzuki cross-coupling of tetrabromothiophene.
  • Palladium-catalyzed addition to nitriles.

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suk-Ku Kang et al.
The Journal of organic chemistry, 61(14), 4720-4724 (1996-07-12)
The palladium-catalyzed cross-coupling reaction of iodinanes (iodonium salts and iodanes) with organoboron compounds to form carbon-carbon bonds was achieved at ambient temperature under aqueous conditions in the absence of base. Coupling of phenylboronic acid with diphenyliodonium tetrafluoroborate in the presence
Robin Frauenlob et al.
The Journal of organic chemistry, 77(9), 4445-4449 (2012-04-13)
We have developed a copper-catalyzed process for the coupling of aldehydes, amines, and boronic acids. This allows greater reactivity with simple aryl boronic acids and allows coupling reactions to proceed that previously failed. Initial mechanistic studies support a process involving
Microwave-enhanced triton B catalyzed Suzuki coupling reaction
Meshram, H. M.; et al.
Indian J. Chem. B, 51, 362-365 (2012)
J J Li et al.
Journal of medicinal chemistry, 39(9), 1846-1856 (1996-04-26)
A novel series of terphenyl methyl sulfones and sulfonamides have been shown to be highly potent and selective cyclooxygenase-2 (COX-2) inhibitors. The sulfonamide analogs 17 and 21 were found to be much more potent COX-2 inhibitors and orally active anti-inflammatory
Yuvraj Satkar et al.
Frontiers in chemistry, 8, 563470-563470 (2020-11-17)
The chemoselective reaction of the C- followed by the O-centered naphthyl radicals with the more electron-deficient hypervalent bond of the diaryliodonium(III) salts is described. This discovered reactivity constitutes a new activation mode of the diaryliodonium(III) salts which enabled a one-pot

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service