Skip to Content
Merck
All Photos(1)

Documents

455059

Sigma-Aldrich

Bisphenol A ethoxylate dimethacrylate

average Mn ~1,700, EO/phenol 15, contains 200 ppm MEHQ as inhibitor

Synonym(s):

2,2-Bis(4-methacryloxypolyethoxyphenyl)propane, Bisphenol A-ethylene oxide adduct dimethacrylate, Ethoxylated bisphenol A dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2(CH2CH2O)nC6H4-4-]2C(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~1,700

contains

200 ppm MEHQ as inhibitor
200 ppm monomethyl ether hydroquinone as inhibitor

composition

EO/phenol, 15

refractive index

n20/D 1.493

density

1.12 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCO.OC(=O)C=C.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2.C3H4O2.C2H6O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;1-2-3(4)5;3-1-2-4/h3-10,16-17H,1-2H3;2H,1H2,(H,4,5);3-4H,1-2H2

InChI key

HGARWRTUWINPJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol A ethoxylate dimethacrylate is an acrylic cross-linker widely used as a precursor to fabricate tissue-mimicking materials for 3D printing, cross-linked gel electrolytes, and to synthesize copolymers via the photopolymerization process.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A study of cross-linked PEO gel polymer electrolytes using bisphenol A ethoxylate diacrylate: ionic conductivity and mechanical properties
Yongku Kang, et al.
Journal of Power Sources, 119-121, 432-437 (2003)
UV light copolymerization of dimethyl vinylphosphonate with bisphenol A ethoxylate dimethacrylate
Lavinia Macarie, et al.
Iranian Polymer Journal, 25, 437-442 (2016)
3D printing of noncytotoxic high-resolution microchannels in Bisphenol-a ethoxylate dimethacrylate tissue-mimicking materials
Roger Domingo-Roca, et al.
3D printing and additive manufacturing (2022)
Sabine H Dickens et al.
Dental materials : official publication of the Academy of Dental Materials, 26(7), 675-681 (2010-04-07)
To establish the relationship of resin composition and resin hydrophilicity (indicated by solubility parameters and logP) to water sorption (WS), solubility, and degree of double bond conversion (DC) of resin mixtures designed for adhesive restoratives by varying the concentration of
Drago Skrtic et al.
Acta biomaterialia, 2(1), 85-94 (2006-05-17)
Improving the anti-demineralizing/remineralizing and mechanical properties of amorphous calcium phosphate (ACP) composites has been the focus of our recent research. In this study, an ethoxylated bisphenol A dimethacrylate (EBPADMA) was blended with triethylene glycol dimethacrylate (TEGDMA), 2-hydroxyethyl methacrylate (HEMA) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service