Skip to Content
Merck
All Photos(2)

Documents

494356

Sigma-Aldrich

Bisphenol A glycerolate dimethacrylate

glycerol/phenol 1

Synonym(s):

2,2′-Bis[4-(3-methacryloxy-2-hydroxypropoxy)phenyl]propane, 2,2′-Bis[4-(3-methacryloyloxy-2-hydroxypropoxy)phenyl]propane, 2,2-Bis[p -(3-methacryloxy-2-hydroxypropoxy)phenyl]propane, 2,2-Bis[p -[2-hydroxy-3-(methacryloxy)propoxy]phenyl]propane, Bisphenol A bis(2-hydroxy-3-methacryloxypropyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2CH2CH(OH)CH2OC6H4-4-]2C(CH3)2
CAS Number:
Molecular Weight:
512.59
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

composition

glycerol/phenol, 1

refractive index

n20/D 1.552 (lit.)

density

1.161 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(O)COc1ccc(cc1)C(C)(C)c2ccc(OCC(O)COC(=O)C(C)=C)cc2

InChI

1S/C29H36O8/c1-19(2)27(32)36-17-23(30)15-34-25-11-7-21(8-12-25)29(5,6)22-9-13-26(14-10-22)35-16-24(31)18-37-28(33)20(3)4/h7-14,23-24,30-31H,1,3,15-18H2,2,4-6H3

InChI key

AMFGWXWBFGVCKG-UHFFFAOYSA-N

General description

Bisphenol A glycerolate dimethacrylate(BisGMA) is an advanced derivative of Bisphenol A(BPA). It is a conventional epoxy methacrylate that is widely used as a monomer in many applications due to the following properties:
  • Phenyl ring provides chemical resistance
  • Ether linkage allows flexibility
  • Hydroxyl group gives good adhesion
  • Methacrylate is the reactive site for crosslinking

Application

Bisphenol A glycerolate dimethacrylate can be used:
  • As a crosslinking monomer in the preparation of zwitter ionic monolithic columns for liquid chromatography.
  • In the preparation of an antibacterial dental adhesive.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Albuquerque Carneiro et al.
The Journal of oral implantology, 40(2), 161-168 (2014-05-02)
The use of provisional resin implant-supported complete dentures is a fast and safe procedure to restore mastication and esthetics of patients soon after surgery and during the adaptation phase to the new denture. This study assessed stress distribution of provisional
Amalia Mazilu Moldovan et al.
Materials (Basel, Switzerland), 12(13) (2019-07-03)
The novelty of this study consists of the formulation and characterization of three experimental bleaching gels with hydroxylapatite oxides and fluorine (G28®, G29®, G30®) based on natural fruit extracts compared to the commercial Opalescence 15% (GC, Ultradent, South Jordan, UT
Bilal Khatri et al.
Micromachines, 11(5) (2020-05-28)
Additive manufacturing, or nowadays more popularly entitled as 3D printing, enables a fast realization of polymer, metal, ceramic or composite devices, which often cannot be fabricated with conventional methods. One critical issue for a continuation of this success story is
Marioara Moldovan et al.
Materials (Basel, Switzerland), 12(13) (2019-07-03)
The novelty of this study consists in the formulation and characterization of three experimental dental composites (PM, P14M, P2S) for cervical dental lesion restoration compared to the commercial composites Enamel plus HRi® - En (Micerium S.p.A, Avengo, Ge, Italy), G-ænial
Wael Aly Ghuneim
The Journal of oral implantology, 39(5), 559-573 (2011-07-20)
This study is a phase of a biomechanical study, a part of a research program concerned with the new concept of in situ tooth replication. The purpose of the study was to evaluate tooth replica under each of two possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service