Skip to Content
Merck
All Photos(3)

Documents

634492

Sigma-Aldrich

4-Pyridinylboronic acid

90%

Synonym(s):

4-Pyridineboronic acid, 4-Pyridylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6BNO2
CAS Number:
Molecular Weight:
122.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

>300 °C (lit.)

storage temp.

−20°C

SMILES string

OB(O)c1ccncc1

InChI

1S/C5H6BNO2/c8-6(9)5-1-3-7-4-2-5/h1-4,8-9H

InChI key

QLULGIRFKAWHOJ-UHFFFAOYSA-N

General description

4-Pyridinylboronic acid is commonly used as a reagent in cross-coupling reactions such as Suzuki-Miyaura cross-coupling.

Application

Reagent used for
  • Palladium-catalyzed Suzuki-Miyaura coupling reactions
  • Ligand-free palladium-catalyzed Suzuki coupling reaction under microwave irradation

Reagent used in Preparation of
  • HIV-1 protease inhibitors
  • Potential cancer threapeutics, such as PDK1 and protein kinase CK2 inhibitors

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligand-free, PdCl2(PPh3)2-catalyzed, microwave-assisted Suzuki coupling of 1-chloro-3-phenylisoquinoline in the synthesis of diversified 1,3-disubstituted isoquinolines
Prabakaran, K.; Nawaz Khan, F.; Jin, J. S.
Research on Chemical Intermediates, 38, 337-346 (2012)
Jin-Tao Yu et al.
Organic & biomolecular chemistry, 10(7), 1359-1364 (2011-12-20)
An efficient palladium-catalyzed Suzuki-Miyaura coupling method involving the reaction between CTV-Br(3) and a variety of aryl and heteroaryl boronic acids in the presence of indolyl phosphane ligands has been developed. This reaction procedure provided a series of C(3)-symmetric aryl-extended rigid
Sumin Lee et al.
Organic letters, 14(9), 2238-2241 (2012-04-28)
The kinetic process of key intermediates involved in the electrochemical ring opening of photochromic dithienylcyclopentenes (DTEs) has been observed for the first time, where the electronic nature of the DTEs is an important factor that determines the rate-determining step in
One-pot approach to N-quinolyl 3?/4?-biaryl carboxamides by microwave-assisted Suzuki--Miyaura coupling and N-boc deprotection
ZY Huang, et al.
The Journal of Organic Chemistry, 81, 9647-9657 (2016)
Jorge Cruz-Huerta et al.
Chemical communications (Cambridge, England), 48(35), 4241-4243 (2012-03-23)
The combination of two heteroaromatic boronic acids with pentaerythritol gave self-complementary tectons which were suitable for the generation of 2D and 3D molecular networks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service