Skip to Content
Merck
All Photos(2)

Key Documents

422479

Sigma-Aldrich

2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol

98%

Synonym(s):

2-(2-Hydroxy-3-tert -butyl-5-methylphenyl)-5-chloro-2H -benzotriazole, 2-(3-tert -Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H -benzotriazole, 2-(5-Chloro-2-benzotriazolyl)-6-tert -butyl-p -cresol, 2-(5-Chloro-2H -benzotriazol-2-yl)-6-(1,1-dimethylethyl)-4-methylphenol, 2-[5-Chloro-2H -benzotriazol-2-yl]-4-methyl-6-(tert -butyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18ClN3O
CAS Number:
Molecular Weight:
315.80
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

144-147 °C (lit.)

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)-n2nc3ccc(Cl)cc3n2

InChI

1S/C17H18ClN3O/c1-10-7-12(17(2,3)4)16(22)15(8-10)21-19-13-6-5-11(18)9-14(13)20-21/h5-9,22H,1-4H3

InChI key

OCWYEMOEOGEQAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol (UV-326) is a benzotriazole based UV absorber that can be used in the photostabilization of industrial materials. It can absorb the harmful UV radiation for the wavelength of about 290 nm.

Application

UV absorber
UV-326 can potentially be used as a UV light filter in cosmetics and sunscreens. It may also be used as a corrosion inhibitor for the protection of building materials and automobiles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behaviour of nanoparticle (ultrafine) titanium dioxide pigments and stabilisers on the photooxidative stability of water based acrylic and isocyanate based acrylic coatings
Allen NS, et al.
Polymer Degradation and Stability, 78(3), 467-478 (2002)
Juzhou Zhang et al.
Se pu = Chinese journal of chromatography, 30(2), 190-195 (2012-06-12)
An effective high performance liquid chromatographic (HPLC) method has been developed for the simultaneous determination of 9 ultraviolet stabilizers in food plastic packaging materials. The food packaging samples were firstly extracted by methanol-ethyl acetate, and then purified by a C18
T H Begley et al.
Food additives and contaminants, 21(10), 1007-1014 (2005-02-17)
The migration characteristics of the UV stabilizer Tinuvin 234 (2-(2H-benzotriazol-2-yl)-4,6-bis (1-methyl-1-phenylethyl)phenol) into food simulants has been measured from polyethylene terephthalate (PET) using HPLC with UV detection. Ethanol/ water, isooctane and a fractionated coconut oil simulant (Miglyol) were used as food
I Carpinteiro et al.
Journal of chromatography. A, 1217(24), 3729-3735 (2010-05-04)
A novel and sensitive method for the determination of five benzotriazole compounds (commonly used as light stabilizers) in indoor dust is presented. Pressurized liquid extraction (PLE) and gas chromatography followed by tandem in time mass spectrometry (GC-MS/MS) were used as
Occurrence and removal of benzotriazoles and ultraviolet filters in a municipal wastewater treatment plant
Liu Y, et al.
Environmental Pollution (Barking, Essex : 1987), 165(9), 225-232 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service