Skip to Content
Merck
All Photos(2)

Key Documents

533203

Sigma-Aldrich

2-(2-Hydroxy-5-methylphenyl)benzotriazole

97%

Synonym(s):

2-(2H-Benzotriazol-2-yl)-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O
CAS Number:
Molecular Weight:
225.25
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

125.5-129.5 °C (lit.)

SMILES string

Cc1ccc(O)c(c1)-n2nc3ccccc3n2

InChI

1S/C13H11N3O/c1-9-6-7-13(17)12(8-9)16-14-10-4-2-3-5-11(10)15-16/h2-8,17H,1H3

InChI key

MCPKSFINULVDNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from an ostomy bag.
E van Hecke et al.
Contact dermatitis, 18(2), 121-122 (1988-02-01)
K Arisu et al.
Contact dermatitis, 26(5), 311-316 (1992-05-01)
We report a 54-year-old female with allergic contact dermatitis from 2-(2-hydroxy-5-methylphenyl)benzotriazole (Tinuvin P) in a spandex tape sewn into underwear. Tinuvin P is used as an ultraviolet light absorber for stabilizing plastics and the other organic materials against discoloration and
B Niklasson et al.
Contact dermatitis, 21(5), 330-334 (1989-11-01)
Contact allergy to the UV-absorber 2-(2-hydroxy-5-methylphenyl)benzotriazole (Tinuvin P) was revealed in a patient with wrist dermatitis caused by a plastic watch strap. A high-performance liquid chromatography method was developed and subsequent chemical analysis showed the presence of Tinuvin P in
Rosa María Claramunt et al.
Molecules (Basel, Switzerland), 12(9), 2201-2214 (2007-10-27)
2-(2,4-Dimethylphenyl)-2H-benzotriazole (1) has been synthesized in a three step procedure starting from 2,4-dimethyl-N-(2-nitrophenyl)benzamide via a 5-(2,4- dimethylphenyl)-1-(2-nitrophenyl)-1H-tetrazole intermediate. Its structure and those of Tinuvin P and 2-phenyl-2H-benzotriazole (5) have been studied by multinuclear NMR (1H-, 13C- and 15N-) in solution
Contact allergy to Tinuvin P.
A C de Groot et al.
Contact dermatitis, 9(4), 324-325 (1983-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service