Skip to Content
Merck
All Photos(2)

Key Documents

535834

Sigma-Aldrich

Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H52N2O4
CAS Number:
Molecular Weight:
480.72
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

loss

0.7 wt. % volatility, 150-175°C

mp

82-85 °C (lit.)

transition temp

Tg >350 °C (dec)

solubility

acetone: 19 % (w/w) at 20 °C
ethyl acetate: 24 % (w/w) at 20 °C
methanol: 38 % (w/w) at 20 °C
chloroform: 45 % (w/w) at 20 °C
methylene chloride: 56 % (w/w) at 20 °C
H2O: insoluble <6 ppm at 20 °C

SMILES string

CC1(C)CC(CC(C)(C)N1)OC(=O)CCCCCCCCC(=O)OC2CC(C)(C)NC(C)(C)C2

InChI

1S/C28H52N2O4/c1-25(2)17-21(18-26(3,4)29-25)33-23(31)15-13-11-9-10-12-14-16-24(32)34-22-19-27(5,6)30-28(7,8)20-22/h21-22,29-30H,9-20H2,1-8H3

InChI key

XITRBUPOXXBIJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2,2,6,6-tetramethyl-4-piperidyl) (T770) sebacate is a low molecular weight ultra violet (UV) hindered amine light stabilizer (HALS) made up of nitroxyl radicals. It acts as a heat controller that can react with phenolic antioxidants incorporated in polymers.

Application

T770 can be used as a hindered amine UV stabilizer in plastic materials. It can be used in the manufacturing and packaging of pharmaceutical products. Titanium oxide(TiO2)/T770 may be used in amine induced light stabilizers in the synthesis of recyclable water-based polyurethane with poly(oxytetramethylene)glycol (PTMG).

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Papke et al.
The Journal of pharmacology and experimental therapeutics, 268(2), 718-726 (1994-02-01)
Bis (2,2,6,6-tetramethyl-4-piperidinyl) sebacate (BTMPS; Tinuvin 770), a sterically hindered amine light and radiation stabilizer manufactured by Ciba-Geigy Corp. (Summit, NJ) and used in a wide range of plastics, inhibits nicotinic acetylcholine receptors expressed in Xenopus oocytes in a use-dependent manner.
Péter Sótonyi et al.
Toxicological sciences : an official journal of the Society of Toxicology, 77(2), 368-374 (2003-12-06)
Tinuvin 770 [bis(2,2,6,6-tetramethyl-4-piperidinyl) sebacate], is a UV light stabilizer plastic additive used worldwide. It is a component of many plastic materials used in medical and food industries. Earlier studies demonstrated its in vitro L-type Ca2+ channel and nicotinic acetylcholine receptor
R Masalha et al.
Brain research, 774(1-2), 260-264 (1998-02-06)
While unproved, environmental toxins of industrial and or agricultural origin represent an attractive theory to explain the increasing incidence of degenerative diseases of the nervous system such as Parkinson's disease (PD). We have examined several chemicals utilized in an area
A molecular study towards the interaction of phenolic anti-oxidants, aromatic amines and HALS stabilizers in a thermo-oxidative ageing process
Barret J, et al.
Polymer Degradation and Stability, 76(3), 441-448 (2002)
V Jackson-Lewis et al.
Brain research, 866(1-2), 197-210 (2000-05-29)
Tinuvin 123, a compound used in the manufacture of plastics, has recently been suggested to possibly cause Parkinson's disease (PD). Herein, we revisited this issue by assessing the effect of Tinuvin 123 on dopaminergic neurons of the substantia nigra following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service