Skip to Content
Merck
All Photos(1)

Key Documents

415464

Sigma-Aldrich

N-Vinylcaprolactam

98%, stabilized

Synonym(s):

1-Ethenylazepan-2-one, 1-Ethenylhexahydro-2H -azepin-2-one, 1-Vinylazepan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO
CAS Number:
Molecular Weight:
139.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

contains

~20 ppm HO-TEMPO as stabilizer

bp

128 °C/21 mmHg (lit.)

mp

35-38 °C (dec.) (lit.)

density

1.029 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CN1CCCCCC1=O

InChI

1S/C8H13NO/c1-2-9-7-5-3-4-6-8(9)10/h2H,1,3-7H2

InChI key

JWYVGKFDLWWQJX-UHFFFAOYSA-N

General description

N-Vinylcaprolactam (NVCL) is an organo-soluble amphiphilic material that is soluble in solvents like benzene, isobutanol, and isopropanol. It can be used in the preparation of poly(NVCL) by free-radical polymerization at a temperature higher than its melting point.

Application

NVCL forms a temperature responsive polymer, which can be used for a variety of biomedical applications.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Liver,Upper respiratory tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (N-vinylcaprolactam), a comprehensive review on a thermoresponsive polymer becoming popular
Cortez-Lemus NA and Licea-Claverie A
Progress in Polymer Science, 53(5), 1-51 (2016)
Synthesis of N-vinylcaprolactam polymers in water-containing media
Lozinsky VI, et al.
Polymer, 41(17), 6507-6518 (2000)
Shane C Halligan et al.
Materials science & engineering. C, Materials for biological applications, 79, 130-139 (2017-06-21)
Poly (N-vinylcaprolactam) (PNVCL) is a polymer which offers superior characteristics for various potential medical device applications. In particular it offers unique thermoresponsive capabilities, which fulfils the material technology constraints required in targeted drug delivery applications. PNVCL phase transitions can be
Gehong Su et al.
Physical chemistry chemical physics : PCCP, 19(40), 27221-27232 (2017-10-05)
In this study, temperature-dependent FTIR spectroscopy in combination with the perturbation-correlation moving-window (PCMW2D) technique and generalized two-dimensional (2D) correlation analysis was applied to investigate the phase transition mechanism of poly(N-vinylcaprolactam) (PVCL) hydrogel upon heating. In the conventional 1D FTIR spectra
Solmaz Banihashem et al.
Carbohydrate polymers, 227, 115333-115333 (2019-10-09)
The gold nanoparticles surface was modified by thioglycolic acid ligand and their surface was coated by the chitosan-grafted-poly(N-vinylcaprolactam) (chitosan-g-PNVCL) copolymer. The cisplatin anticancer drug was loaded into the synthesized nanocarriers and its performance was investigated for the treatment of MCF-7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service