Skip to Content
Merck
All Photos(2)

Key Documents

440914

Sigma-Aldrich

2,2′-Azobis(2-methylpropionamidine) dihydrochloride

powder or granules, 97%

Synonym(s):

α,α′-Azodiisobutyramidine dihydrochloride, AAPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[=NC(CH3)2C(=NH)NH2]2·2HCl
CAS Number:
Molecular Weight:
271.19
Beilstein:
3718854
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder or granules

t1/2

10 hr(56 °C)

mp

175-177 °C (lit.)

solubility

acetone, dioxane, methanol, ethanol, DMSO and water: soluble

SMILES string

Cl.Cl.CC(C)(\N=N\C(C)(C)C(N)=N)C(N)=N

InChI

1S/C8H18N6.2ClH/c1-7(2,5(9)10)13-14-8(3,4)6(11)12;;/h1-4H3,(H3,9,10)(H3,11,12);2*1H/b14-13+;;

InChI key

LXEKPEMOWBOYRF-QDBORUFSSA-N

Application

Free radical initiator.
Polymerization initiator for acrylic, vinyl and allyl monomers.

Features and Benefits

Undergoes first order decomposition to a cationic radical. Decomposes on exposure to UV. Compatible with cationic surfactants. Decomposition rate is pH dependent

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Self-heat. 1 - Skin Sens. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Yu Wang et al.
Biomacromolecules, 13(9), 2616-2623 (2012-07-20)
PEO-PPO-PEO triblock copolymers have opposing effects on lipid membrane integrity: they can behave either as membrane sealants or as membrane permeabilizers. To gain insights into their biomembrane activities, the fundamental interactions between a series of PEO-based polymers and phospholipid vesicles
Nitya Jani et al.
Journal of pharmacological and toxicological methods, 65(3), 142-146 (2012-04-18)
Radical-induced haemolysis has been employed by many investigators to determine the antioxidant capacity of novel compounds. However the free radical generator 2,2'-azobis (2-amidinopropane) dihydrochloride (AAPH) results in the complete depletion of intracellular reduced glutathione (GSH) in cells that can no
Chuan Xiao et al.
European journal of medicinal chemistry, 53, 159-167 (2012-04-24)
A series of 4-methylcoumarin derivatives containing 4,5-dihydropyrazole moiety were synthesized and their antioxidant activities were evaluated in AAPH (2,2'-azobis(2-amidinopropane hydrochloride))-induced oxidation of DNA, and in trapping DPPH (2,2'-diphenyl-1-picrylhydrazyl) and ABTS(+•) (2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cationic radical), respectively. Among coumarin derivatives, 3a-d and 4a-c
Shawn N Watson et al.
Neurobiology of aging, 34(2), 610-620 (2012-03-31)
Memory failure associated with changes in neuronal circuit functions rather than cell death is a common feature of normal aging in diverse animal species. The (neuro)biological foundations of this phenomenon are not well understood although oxidative stress, particularly in the
A Nakajima et al.
Analytical and bioanalytical chemistry, 403(7), 1961-1970 (2012-05-01)
The characteristics of the spin-trapping reaction in the oxygen radical absorbance capacity (ORAC)-electron spin resonance (ESR) assay were examined, focusing on the kind of spin traps. 2,2-Azobis(2-amidinopropane) dihydrochloride (AAPH) was used as a free radical initiator. The spin adducts of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service