Skip to Content
Merck
All Photos(4)

Key Documents

C2204

Sigma-Aldrich

ε-Caprolactam

99%

Synonym(s):

epsilon-Caprolactam, 2-Oxohexamethyleneimine, Aza-2-cycloheptanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
106934
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

99%

form

crystals

autoignition temp.

707 °F

expl. lim.

8 %

bp

136-138 °C/10 mmHg (lit.)

mp

68-71 °C (lit.)

SMILES string

O=C1CCCCCN1

InChI

1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)

InChI key

JBKVHLHDHHXQEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ε-Caprolactam can be used as a precursor for the production of nylon-6 by ring-opening polymerization.[1][2]It is also microwave irradiated with ε caprolactone in the presence of an anionic catalyst to yield poly(ε caprolactam-co-ε-caprolactone).[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of a ?green? one-step catalytic production of ?-caprolactam (precursor of nylon-6)
Thomas JM and Raja R
Proceedings of the National Academy of Sciences of the USA, 102(39), 13732-13736 (2005)
Microwave syntheses of poly (?-caprolactam-co-?-caprolactone)
Fang X, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 38(8), 1379-1390 (2000)
?-Caprolactam: New by-product free synthesis routes
Dahlhoff, G, et al.
Catalysis Reviews: Science and Engineering, 43(4), 381-441 (2001)
Ahmad Bitar et al.
Journal of biomedical nanotechnology, 8(5), 709-719 (2012-08-15)
Temperature and glucose sensitive microgels were prepared via radical precipitation polymerization, using N-vinylcaprolactam (NVCL) as the main monomer, N,N-methylenebisacrylamide (MBA) as a crosslinker, V50 as initiator and 4-vinylphenylboronic acid (VPBA) as a functional monomer. The LCST of homopoly (NVCL) was
Georgeta Voicu et al.
International journal of pharmaceutics, 446(1-2), 63-69 (2013-02-14)
Here, we report the fabrication of a novel ε-caprolactam-silica (ε-SiO2) network and assessed its biocompatibility and ability to improve the antimicrobial activity of kanamycin. The results of the quantitative antimicrobial assay demonstrate that the obtained ε-SiO2 network has efficiently improved

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service