Skip to Content
Merck
All Photos(2)

Key Documents

414891

Sigma-Aldrich

Cadmium

granular, 30-80 mesh, ≥99%

Synonym(s):

Cd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

granular

reaction suitability

core: cadmium
reagent type: catalyst

resistivity

7.27 μΩ-cm, 22°C

particle size

30-80 mesh

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cadmium is a naturally occurring element found in the earth′s crust. Usually, it is found as a mineral combined with oxygen (cadmium oxide), chlorine (cadmium chloride), or sulfur (cadmium sulfate, cadmium sulfide). Cadmium is a relatively rare element that is not found in its pure form in nature. It is possible to combine cadmium with several nonferrous metals to form alloys with useful commercial properties. The five main applications of cadmium are:
  • nickel-cadmium battery electrodes
  • plastic, ceramic, and glass pigments
  • polyvinyl chloride (PVC) stabilization against light and heat
  • engineering coatings for steel and other nonferrous metals
  • a component of several specialized alloys.

Application

Cadmium granules is used to reduce nitrate to nitrite.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New electrochemical sensors for detection of nitrites and nitrates
Badea M, et al.
Journal of Electroanalytical Chemistry, 509(1), 66-72 (2001)
Boldizsar Nagy et al.
Acta chimica Slovenica, 60(2), 263-273 (2013-07-25)
Biosorption of cadmium ions from synthetic aqueous solution using popular Romanian fir tree sawdust (Abies Alba) as biosorbent, was investigated in this work. Prior to its utilization the considered biomass was washed, dried and sieved without further chemical treatments. The
F Tomei et al.
Annali di igiene : medicina preventiva e di comunita, 25(4), 353-363 (2013-05-25)
The exposure to Cd has been linked to many disease and liver is the major target organ of Cd toxicity following acute or chronic exposure.No study has ever evaluated the association between liver function tests and exposure to Cd present
Gang Zhang et al.
Journal of environmental sciences (China), 25(1), 231-235 (2013-04-17)
The emission characteristics of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs) and heavy metals were evaluated during co-combustion of sewage sludge with coal from a circulating fluidized bed incinerator. The stack gas, slag and fly ash samples were sampled and analyzed. The
Simona Ben-Haim et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 873-879 (2013-04-13)
Myocardial perfusion imaging (MPI) is well established in the diagnosis and workup of patients with known or suspected coronary artery disease (CAD); however, it can underestimate the extent of obstructive CAD. Quantification of myocardial perfusion reserve with PET can assist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service