Skip to Content
Merck
All Photos(3)

Documents

435236

Sigma-Aldrich

Cellulose

microcrystalline, powder

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

microcrystalline
powder

particle size

51 μm

pH

5-7 (11 wt. %)

bulk density

0.6 g/mL (25°C)

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bulking agent, opacifier, anti-caking agent, extrusion aid and stabilizer for foams and emulsions.
High purity cellulose powders for partition chromatography.

Features and Benefits

Amorphous regions are hydrolyzed leaving crystalline microfibrils. Forms thixotropic gels, good thermal stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoang T M D Le et al.
PloS one, 16(2), e0247076-e0247076 (2021-02-12)
The effects of nutrient and mechanical sensing on gut motility and intestinal metabolism in lower vertebrates remains largely unknown. Here we present the transcriptome response to luminal stimulation by nutrients and an inert bolus on nutrient response pathways and also
Eshchar Mizrachi et al.
The New phytologist, 194(1), 54-62 (2012-04-05)
Fast-growing, short-rotation forest trees, such as Populus and Eucalyptus, produce large amounts of cellulose-rich biomass that could be utilized for bioenergy and biopolymer production. Major obstacles need to be overcome before the deployment of these genera as energy crops, including
Nathalie Lavoine et al.
Carbohydrate polymers, 90(2), 735-764 (2012-07-31)
Interest in microfibrillated cellulose (MFC) has been increasing exponentially. During the last decade, this bio-based nanomaterial was essentially used in nanocomposites for its reinforcement property. Its nano-scale dimensions and its ability to form a strong entangled nanoporous network, however, have
Okako Omadjela et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(44), 17856-17861 (2013-10-16)
Cellulose is a linear extracellular polysaccharide. It is synthesized by membrane-embedded glycosyltransferases that processively polymerize UDP-activated glucose. Polymer synthesis is coupled to membrane translocation through a channel formed by the cellulose synthase. Although eukaryotic cellulose synthases function in macromolecular complexes
Inka Vanwonterghem et al.
The ISME journal, 8(10), 2015-2028 (2014-04-18)
A replicate long-term experiment was conducted using anaerobic digestion (AD) as a model process to determine the relative role of niche and neutral theory on microbial community assembly, and to link community dynamics to system performance. AD is performed by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service