Skip to Content
Merck
All Photos(2)

Key Documents

265330

Sigma-Aldrich

Cadmium

granular, ≥99%, 5-20 mesh

Synonym(s):

Cd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

granular

reaction suitability

core: cadmium
reagent type: catalyst

resistivity

7.27 μΩ-cm, 22°C

particle size

5-20 mesh

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David A Kimberly et al.
Environmental pollution (Barking, Essex : 1987), 184, 419-425 (2013-10-15)
Latent effects result from embryonic experiences but manifest in later stages of ontogeny. Our objective was to better understand how developmental exposure to contaminants influence life history traits and tolerance to novel stress in the freshwater gastropod, Physa pomilia. Ten
T C Robson et al.
Environmental pollution (Barking, Essex : 1987), 184, 283-289 (2013-10-01)
The biogeochemistry and bioavailability of cadmium, released during sphalerite weathering in soils, were investigated under contrasting agricultural scenarios to assess health risks associated with sphalerite dust transport to productive soils from mining. Laboratory experiments (365 d) on temperate and sub-tropical soils
Simona Ben-Haim et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 873-879 (2013-04-13)
Myocardial perfusion imaging (MPI) is well established in the diagnosis and workup of patients with known or suspected coronary artery disease (CAD); however, it can underestimate the extent of obstructive CAD. Quantification of myocardial perfusion reserve with PET can assist
Bettina Julin et al.
Epidemiology (Cambridge, Mass.), 24(6), 880-885 (2013-09-14)
The toxic metal cadmium is suggested to increase the risk of cardiovascular disease, but only one incidence study has explored this association. We evaluated the association between quartiles of food frequency questionnaire-based estimates of cadmium exposure from food (the predominant
Renee M Gardner et al.
American journal of epidemiology, 177(12), 1356-1367 (2013-05-17)
In this prospective cohort study, based on 1,505 mother-infant pairs in rural Bangladesh, we evaluated the associations between early-life exposure to arsenic, cadmium, and lead, assessed via concentrations in maternal and child urine, and children's weights and heights up to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service