Skip to Content
Merck
All Photos(2)

Key Documents

410497

Sigma-Aldrich

4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran

Dye content 98 %

Synonym(s):

DCM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3O
CAS Number:
Molecular Weight:
303.36
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

composition

Dye content, 98%

mp

215-220 °C (lit.)

λmax

468 nm

OLED Device Performance

ITO/Alq3:DCM/Alq3/Mg:Ag

  • Color: red
  • Max. EQE: 1.3 %

ITO/TPD/Alq3:DCM (10%)/Alq3/Mg:Ag
  • Color: red
  • Max. Luminance: 150 Cd/m2

SMILES string

CN(C)c1ccc(\C=C\C2=CC(\C=C(C)O2)=C(\C#N)C#N)cc1

InChI

1S/C19H17N3O/c1-14-10-16(17(12-20)13-21)11-19(23-14)9-6-15-4-7-18(8-5-15)22(2)3/h4-11H,1-3H3/b9-6+

InChI key

YLYPIBBGWLKELC-RMKNXTFCSA-N

General description

4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran (DCM) is a red laser dye that consists of dicyanomethylene as an electron acceptor and dimethylaniline group as an electron donor. It has a π-conjugated 4H-pyran-4-ylidiene which bridges both the acceptor/donor groups. It can be used as a dopant and also in organic solid state lasers.

Application

DCM can be used as a laser dye to enhance the emission of distributed feedback (DFB) device by FÖrster resonance energy transfer (FRET). It may be used as a capping layer that allows the conversion of blue to red colored emission in organic light emitting diodes (OLED). DCM may also find potential applications in the enhancement of energy transfer of different devices like metal organic frameworks (MOFs), dye sensitized solar cells (DSSCs) and polarity sensors.

Features and Benefits

Voltage-tunable multicolor emission with enhanced luminance (~1000 cd/m2) was observed using varying amounts of DCM dye in a polymer light-emitting diode (PLED).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tunable two-color luminescence and host-guest energy transfer of fluorescent chromophores encapsulated in metal-organic frameworks.
Yan D, et al.
Scientific reports, 4(3), 4337-4337 (2014)
Solid state solvation effect and reduced amplified spontaneous emission threshold value of glass forming DCM derivative in PMMA films.
Vembris A, et al.
Journal of Luminescence, 158(7), 441-446 (2015)
Ashim Pramanik et al.
Journal of colloid and interface science, 565, 326-336 (2020-01-25)
The development of white light-emitting LED (WLED) is now one of the important demands of present society due to its energy saving feature. However, in this work, an ingenious strategy has been employed to develop Forster resonance energy transfer (FRET)
Arnab Halder et al.
Langmuir : the ACS journal of surfaces and colloids, 20(3), 653-657 (2005-03-19)
Solvation dynamics of 4-(dicyanomethylidene)-2-[p-(dimethylamino)styryl]-6-methyl-4H-pyran (DCM) is studied in a polypeptide-surfactant aggregate consisting of gelatin and sodium dodecyl sulfate (SDS) in potassium dihydrogen phosphate (KP) buffer. The average solvation time (tauS) in gelatin-SDS aggregate at 45 degrees C is found to
Tunable organic distributed feedback dye laser device excited through Forster mechanism.
Tsutsumi N and Hinode T
Applied Physics B: Lasers and Optics, 123(3), 93-93 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service