Skip to Content
Merck
All Photos(1)

Key Documents

76509

Supelco

Pentadecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH3
CAS Number:
Molecular Weight:
212.41
Beilstein:
1698194
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

7.4 (vs air)

vapor pressure

1 mmHg ( 91.6 °C)

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.431 (lit.)
n20/D 1.431

bp

270 °C (lit.)

mp

8-10 °C (lit.)

transition temp

solidification point 8.5-10 °C

density

0.769 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCC

InChI

1S/C15H32/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3-15H2,1-2H3

InChI key

YCOZIPAWZNQLMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentadecane is a hydrocarbon, which is found to be highly viscous in nature.[1][2][3] It is formed as a decomposition product of glycidyl palmitate of fatty acid, when subjected to heating in the temperature range of 180-200°C.[3]
Pentadecane, belonging to the class of aliphatic hydrocarbons,[4] is identified as a volatile flavor component in roasted pork of Mini-pig,[5] Yerba mate,[4] Milano salami,[6] virgin high density polyethylene (HDPE), etc.[7] It plays an important role as a chemical intermediate for producing a variety of products, solvents and as a component of gasoline and diesel fuel.[8] Pentadecane is also determined as an aroma component in unifloral Greek citrus honey,[9] Niva cheese,[10] cooked black rice, etc.[11]

Application

Pentadecane has been used as an analytical reference standard for the quantification of the analyte in gasoline samples using gas chromatography technique. It may be used as an analytical reference standard for the quantification of the analyte in:
  • Roasted pork of Mini-pig using gas chromatography and mass spectrometry (GC–MS).[5]
  • Yerba mate using gas chromatography–mass spectrometry (GC–MS) and gas chromatography–olfactometry (GC–O).[4]
  • Milano salami using gas chromatography with mass spectrometry (GC-MS), flame ionization detection (GC-FID) and olfactometry analysis (GC-O).[6]

Pentadecane may be used as a phase change material, in combination with capric acid and lauric acid, and their thermal behaviour can be studied by using differential scanning colorimetry (DSC).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compounds of commercial Milano salami
Meynier A, et al.
Meat Science, 51(2), 175-183 (1999)
Characterization of volatile aroma compounds in cooked black rice
Yang SD, et al.
Journal of Agricultural and Food Chemistry, 56(1), 235-240 (2008)
Agarwal R et al.
The Ultimate BMAT Guide: 800 Questions (2017)
Volatile flavor constituents in roasted pork of Mini-pig
Xie J, et al.
Food Chemistry, 109(3), 506-514 (2008)
Decomposition products of glycidyl esters of fatty acids by heating
Kimura W and Endo Y
Bioscience, Biotechnology, and Biochemistry, 81(3), 581-586 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service