Skip to Content
Merck
All Photos(3)

Key Documents

B65551

Sigma-Aldrich

1-Bromododecane

97%

Synonym(s):

Dodecyl bromide, Lauryl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11Br
CAS Number:
Molecular Weight:
249.23
Beilstein:
506159
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.6 (vs air)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

134-135 °C/6 mmHg (lit.)

mp

−11-−9 °C (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCBr

InChI

1S/C12H25Br/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-12H2,1H3

InChI key

PBLNBZIONSLZBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Bromododecane can be used for modifying the solubility and processability by alkylation. Some of the reported applications include:
  • Improving the solubility of rigid polycyclic aromatic hydrocarbons used in organic electronics to ease the solution processability.
  • Inducing hydrophobicity ethoxylated urethanes and polyurethanes.
  • Increasing lipophilicity of molecules for biological applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contorted polycyclic aromatic hydrocarbons with cove regions and zig-zag edges.
Chen Y, et al.
Chemical Communications (Cambridge, England), 53(60), 8474-8477 (2017)
Double-tailed long chain BODIPYs-Synthesis, characterization and preliminary studies on their use as lipid fluorescence probes.
Arranja CT, et al.
Journal of Molecular Structure, 1146, 62-69 (2017)
Solution-processable 2,1,3-benzothiadiazole containing compound based on the novel 1-dodecyl-6-dodecoxynaphthyridine-2-one unit for organic field-effect transistors.
Cameron J, et al.
Organic Electronics, 49, 400-405 (2017)
Rinku Dutta et al.
Pharmaceutical research, 34(3), 564-578 (2016-12-21)
To determine the pharmacokinetic parameters and biodistribution of GDC-0449 loaded polymeric micelles after systemic administration into common bile duct ligation (CBDL) induced liver fibrotic mice. We used GDC-0449 encapsulated methoxy poly (ethylene glycol)-block-poly (2-methyl-2-carboxyl-propylene carbonate)-graft-dodecanol (PEG-PCD) non-targeted polymeric micelles for
Synthesis and Properties of New N?Heteroheptacenes for Solution?Based Organic Field Effect Transistors.
Zhou F, et al.
Chemistry?A European Journal , 23(51), 12542-12549 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service