Skip to Content
Merck
All Photos(1)

Key Documents

546186

Sigma-Aldrich

Coumarin 153

Dye content 99 %

Synonym(s):

2,3,6,7-Tetrahydro-9-(trifluoromethyl)-1H,5H,11H-[1]benzopyrano(6,7,8-ij)quinolizin-11-one, 2,3,6,7-Tetrahydro-9-trifluoromethyl-1H,5H-quinolizino(9,1-gh)coumarin, 8-Trifluoromethyl-2,3,5,6-4H-1,H-11-oxa-3a-aza-benzo[de]anthracen-10-one, Coumarin 540A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3NO2
CAS Number:
Molecular Weight:
309.28
Beilstein:
3624201
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

composition

Dye content, 99%

mp

164-168 °C (lit.)

λmax

422 nm (lit.)
422 nm

fluorescence

λem 532 nm in ethanol (Lasing peak 550 nm, lasing range 521 - 605 nm (methanol), pump source XeCl (308 nm))

SMILES string

FC(F)(F)C1=CC(=O)Oc2c3CCCN4CCCc(cc12)c34

InChI

1S/C16H14F3NO2/c17-16(18,19)12-8-13(21)22-15-10-4-2-6-20-5-1-3-9(14(10)20)7-11(12)15/h7-8H,1-6H2

InChI key

VSSSHNJONFTXHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Laser dye

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam H Turner et al.
The Journal of chemical physics, 149(17), 174503-174503 (2018-11-10)
The equilibrium and dynamic solvation responses of coumarin 153 (C153) in a range of deep eutectic solvents (DESs) based on choline chloride with either urea (molar ratio 1:2, ChCl:U), glycerol (1:2, ChCl:G), ethylene glycol (1:2, ChCl:E), or malonic acid (1:1
Lien D'Olieslaeger et al.
Journal of colloid and interface science, 504, 527-537 (2017-06-13)
Conjugated polymers are versatile bio-imaging probes as their optical properties can be readily fine-tuned. In this manuscript, fluorescent conjugated polymer nanoparticles are fabricated using three different poly(p-phenylene ethynylene) (PPE) derivatives. The polymers have the same backbone but carry different side
Dilek Yalcin et al.
Physical chemistry chemical physics : PCCP, 22(1), 114-128 (2019-12-10)
Ionic liquids (ILs) are highly tailorable solvents with many potential applications. Knowledge about their solvation properties is highly beneficial in the utilization of ILs for specific tasks, though for many ILs this is currently unknown. In this study, we have
Vitaliy Kapishon et al.
Biomacromolecules, 16(7), 2040-2048 (2015-06-13)
Alginate-based amphiphilic graft copolymers were synthesized by single electron transfer living radical polymerization (SET-LRP), forming stable micelles during polymerization induced self-assembly (PISA). First, alginate macroinitiator was prepared by partial depolymerization of native alginate, solubility modification and attachment of initiator. Depolymerized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service