Skip to Content
Merck
All Photos(2)

Documents

394459

Sigma-Aldrich

Perfluorononanoic acid

97%

Synonym(s):

Heptadecafluorononanoic acid, Perfluorononanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7COOH
CAS Number:
Molecular Weight:
464.08
Beilstein:
1897287
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

59-62 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C9HF17O2/c10-2(11,1(27)28)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)26/h(H,27,28)

InChI key

UZUFPBIDKMEQEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuemei Fang et al.
Toxicological sciences : an official journal of the Society of Toxicology, 108(2), 367-376 (2009-02-07)
Perfluorononanoate (PFNA), a perfluorinated alkyl acid containing nine carbon chains, has been detected in abiotic and biotic matrices worldwide. Although a few studies have reported toxic effects of PFNA, little information of the mechanism has been offered. In this study
Marleen Maras et al.
Environmental health perspectives, 114(1), 100-105 (2006-01-06)
We investigated estrogen-like properties of five perfluorinated compounds using a combination of three in vitro assays. By means of an E-screen assay, we detected the proliferation-promoting capacity of the fluorotelomer alcohols 1H,1H,2H,2H-perfluorooctan-1-ol (6:2 FTOH) and 1H,1H,2H,2H-perfluoro-decan-1-ol (8:2 FTOH). The more
Xuemei Fang et al.
Toxicology, 294(2-3), 109-115 (2012-03-06)
Exposure to perfluorononanoic acid (PFNA), an increasingly persistent organic pollutant that has been detected in abiotic and biotic matrices, has been demonstrated to cause hepatotoxicity in animals. However, the effects of PFNA on hepatic glucose metabolism have not been fully
Katoria Tatum-Gibbs et al.
Toxicology, 281(1-3), 48-55 (2011-01-18)
Perfluorononanoic acid (PFNA) is a fluorinated organic chemical found at low levels in the environment, but is detectable in humans and wildlife. The present study compared the pharmacokinetic properties of PFNA in two laboratory rodent species. Male and female Sprague-Dawley
Lin Tao et al.
Environmental science & technology, 42(9), 3472-3478 (2008-06-05)
The collapse of the World Trade Center (WTC) on September 11, 2001 resulted in the release of several airborne pollutants in and around the site. Perfluorochemicals including perfluorooctanesulfonate (PFOS) and perfluorooctanoic acid (PFOA), which are used in soil- and stain-resistant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service