Skip to Content
Merck
All Photos(1)

Documents

80444

Supelco

Perfluoroundecanoic acid

analytical standard

Synonym(s):

PFAS, Heneicosafluoroundecanoic acid, PFUnDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)9CO2H
CAS Number:
Molecular Weight:
564.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

Assay

≥96.0% (GC)
96.0-104.0% (T)

shelf life

limited shelf life, expiry date on the label

bp

160 °C/60 mmHg (lit.)

mp

96-101 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C11HF21O2/c12-2(13,1(33)34)3(14,15)4(16,17)5(18,19)6(20,21)7(22,23)8(24,25)9(26,27)10(28,29)11(30,31)32/h(H,33,34)

InChI key

SIDINRCMMRKXGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perfluoroundecanoic acid belongs to the class of persistent perfluorinated compounds, found in environmental and biological samples. They are used in various commercial applications such as in firefighting foams, detergents, insecticides, paper, food containers as well as to produce photographic films and manufacture fat- and water-resistant fabrics, etc.

Application

Perfluorotetradecanoic acid may be used as an analytical reference standard for the quantification of the analyte in water samples using a high-performance liquid chromatography technique.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of perfluoroalkyl acids and sulphonates in Italian eel samples by HPLC-HRMS Orbitrap
Chiesa ML, et al.
Chemosphere, 193, 358-364 (2018)
Perfluorinated compounds in the Pearl river and Yangtze river of China
So KM, et al.
Chemosphere, 68(11), 2085-2095 (2007)
HPLC determination of perfluorinated carboxylic acids with fluorescence detection
Pobozy E, et al.
Microchimica Acta, 172(3-4), 409-417 (2011)
Luiz Gustavo Dos Anjos Borges et al.
Viruses, 12(6) (2020-07-02)
Live visualization of influenza A virus (IAV) structural proteins during viral infection in cells is highly sought objective to study different aspects of the viral replication cycle. To achieve this, we engineered an IAV to express a Tetra Cysteine tag
Monika Stróżyńska et al.
European journal of mass spectrometry (Chichester, England), 26(2), 131-143 (2019-10-09)
A structural investigation of perfluorocarboxylic acid derivatives formed in the reaction with N,N-dimethylformamide dialkylacetals employing several techniques of mass spectrometry (MS) is described. Two derivatizing reagents, dimethylformamide dimethyl acetal (DMF-DMA) and dimethylformamide diethylacetal (DMF-DEA) were used. In contrast to carboxylic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service