Skip to Content
Merck
All Photos(1)

Key Documents

33607

Supelco

Heptadecafluorooctanesulfonic acid solution

100 μg/mL in methanol, analytical standard

Synonym(s):

PFAS, PFOS, Perfluorooctanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7SO3H
CAS Number:
Molecular Weight:
500.13
Beilstein:
1813859
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Agency

suitable for ISO 21675 2019
suitable for ISO 25101

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

PFAS testing
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8HF17O3S/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28/h(H,26,27,28)

InChI key

YFSUTJLHUFNCNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptadecafluorooctanesulfonic acid belongs to the class of perfluorinated alkyl acids and is emerging as persistent organic contaminants in the environment.

Application

Heptadecafluorooctanesulfonic acid may be used as an analytical reference standard for the determination of the analyte in water samples using solid-phase microextraction combined with high-performance liquid chromatography with electrospray-ion trap mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing, or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass. The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent. After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley V Schwartz et al.
Development (Cambridge, England), 148(9) (2021-05-08)
Optimal embryonic development plays a major role in the health of an individual beyond the developmental stage. Nutritional perturbation during development is associated with cardiovascular and metabolic disease later in life. With both nutritional uptake and overall growth being risk
Comparison of human whole blood, plasma, and serum matrices for the determination of perfluorooctanesulfonate (PFOS), perfluorooctanoate (PFOA), and other fluorochemicals.
Ehresman JD, et al.
Environmental Research, 103(2), 176-184 (2007)
Michio Murakami et al.
Chemosphere, 93(1), 140-145 (2013-06-12)
The formation of perfluorinated surfactants (PFSs) from their precursors in waters is of concern. In this study, the formation of PFSs through biodegradation of precursors was measured in incubation tests. Indigenous microorganisms in groundwater were able to biodegrade perfluorooctane sulfonamide
Determination of perfluorooctanesulfonate and perfluorooctanoate in water samples by SPE-HPLC/electrospray ion trap mass spectrometry.
Teng J, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 93(1), 55-59 (2009)
Henrik Viberg et al.
Toxicology, 304, 185-191 (2013-01-05)
Perfluoroalkyl acids, including perfluorohexane sulfonate (PFHxS), are fluorinated organic compounds used as surfactants and water and stain repellents in carpets, paper, and textiles, with characteristics to bioaccumulate and biomagnify in the food chain. PFHxS is found in umbilical cord blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service